コンテンツへスキップ
Merck
  • Vibrational spectroscopy of the mineral meyerhofferite CaB3O3(OH)5·H2O--an assessment of the molecular structure.
Vibrational spectroscopy of the mineral meyerhofferite CaB3O3(OH)5·H2O--an assessment of the molecular structure.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-06-12)
Ray L Frost, Andrés López, Yunfei Xi, Ricardo Scholz, Geraldo Magela da Costa, Fernanda M Belotti, Rosa Malena Fernandes Lima
要旨
Meyerhofferite is a calcium hydrated borate mineral with ideal formula: CaB3O3(OH)5·H2O and occurs as white complex acicular to crude crystals with length up to ~4 cm, in fibrous divergent, radiating aggregates or reticulated and is often found in sedimentary or lake-bed borate deposits. The Raman spectrum of meyerhofferite is dominated by intense sharp band at 880 cm(-1) assigned to the symmetric stretching mode of trigonal boron. Broad Raman bands at 1046, 1110, 1135 and 1201 cm(-1) are attributed to BOH in-plane bending modes. Raman bands in the 900-1000 cm(-1) spectral region are assigned to the antisymmetric stretching of tetrahedral boron. Distinct OH stretching Raman bands are observed at 3400, 3483 and 3608 cm(-1). The mineral meyerhofferite has a distinct Raman spectrum which is different from the spectrum of other borate minerals, making Raman spectroscopy a very useful tool for the detection of meyerhofferite in sedimentary and lake bed deposits.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
Calcium metaborate, technical, 39-44% B2O3 basis, 31-37% CaO basis, powder