コンテンツへスキップ
Merck
  • Solvent effect on the construction of two microporous yttrium-organic frameworks with high thermostability via in situ ligand hydrolysis.
Solvent effect on the construction of two microporous yttrium-organic frameworks with high thermostability via in situ ligand hydrolysis.
Dalton transactions (Cambridge, England : 2003) (2010-05-21)
Bao-Xia Dong, Xiao-Jun Gu, Qiang Xu
要旨
Two novel yttrium trimesates with open frameworks have been synthesized by the self-assembly of trimethyl 1,3,5-benzenetricarboxylate and Y(3+) ion in mixed solvent of dimethylformamide (DMF)/water and diethylformamide (DEF)/water, where changes of solvents with different molecular sizes lead to the formation of two similar three-dimensional frameworks with different spatial arrangement styles of the linkers around the Y(3+) centers, and solvent-dependent one-dimensional channels (ca. 6.2 x 7.7 A(2)) along the [100] direction, as shown by X-ray crystallography. Structural analyses indicate that they have excellent thermal stabilities which are endurable up to 490 degrees C.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
トリメチル 1,3,5-ベンゼントリカルボキシラート, 98%