コンテンツへスキップ
Merck
  • Inhibitory effects of the methanol extract of Ganoderma lucidum on mosquito allergy-induced itch-associated responses in mice.
Inhibitory effects of the methanol extract of Ganoderma lucidum on mosquito allergy-induced itch-associated responses in mice.
Journal of pharmacological sciences (2010-10-16)
Tsugunobu Andoh, Qun Zhang, Takumi Yamamoto, Manabu Tayama, Masao Hattori, Ken Tanaka, Yasushi Kuraishi
要旨
Recently, we showed that a methanol extract of Ganoderma lucidum inhibits scratching, an itch-related response, induced by intradermal injections of some pruritogens in mice. The present study investigated whether G. lucidum extract would inhibit allergic itch. In mice sensitized with an extract of salivary gland of mosquito (ESGM), an intradermal injection of ESGM elicited scratching, which was suppressed by oral administration of G. lucidum extract (100 and 300 mg/kg). The scratching was inhibited by the H₁ histamine-receptor antagonist azelastine, but not by the peripherally acting H₁-antagonist terfenadine, at the oral dose of 30 mg/kg. In sensitized mice, ESGM increased the activity of cutaneous nerve, which was suppressed by G. lucidum extract (300 mg/kg). Although terfenadine (30 mg/kg) inhibited plasma extravasation induced by ESGM in the sensitized mice, G. lucidum extract (300 mg/kg) was without effect. These results suggest that G. lucidum extract relieves allergic itch through a peripheral action. The results support the idea that mast cells and H₁ histamine receptors are not the primary sites of the antipruritic action of G. lucidum extract.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
アゼラスチン 塩酸塩, ≥98% (HPLC)
アゼラスチン 塩酸塩, European Pharmacopoeia (EP) Reference Standard