コンテンツへスキップ
Merck
  • Nitric oxide-dependent and -independent mechanisms are involved in TNF-alpha -induced depression of cardiac myocyte contractility.
Nitric oxide-dependent and -independent mechanisms are involved in TNF-alpha -induced depression of cardiac myocyte contractility.
American journal of physiology. Regulatory, integrative and comparative physiology (2007-01-20)
Anand Kumar, Bhanu Paladugu, Joel Mensing, Aseem Kumar, Joseph E Parrillo
要旨
Previous studies have demonstrated the presence of myocardial depression in clinical and experimental septic shock. This response is mediated, in part, through circulating TNF-alpha-induced, nitric oxide-dependent, depression of basal myocyte contractility. Other mechanisms of early myocardial dysfunction involving decreased response to adrenergic stimulation may exist. This study evaluated the presence and nitric oxide dependence of impaired adrenergic response to TNF-alpha in in vitro cardiac myocytes. The contraction of electrically paced neonatal rat cardiac myocytes in tissue culture was quantified using a closed-loop video tracking system. TNF-alpha induced depression of baseline contractility over the first 20 min of cardiac myocyte exposure. This effect was blocked by N-methyl-arginine (NMA), a nitric oxide synthase inhibitor, in all studies. Contractile and cAMP response to increasing concentrations of isoproterenol was deficient in cardiac myocytes exposed to TNF-alpha regardless of the presence of NMA. In contrast, increasing concentrations of forskolin (a direct stimulant of adenylate cyclase) and dibutyryl cAMP (a metabolically active membrane-soluble analog of cAMP) completely reversed TNF-alpha-mediated depression, though only in the presence of NMA. Forskolin-stimulated cAMP generation remained intact regardless of NMA. Increasing concentrations of exogenous calcium chloride, unlike other inotropic agents, corrected TNF-alpha-mediated defects of contractility independent of the presence of NMA. These data suggest that TNF-alpha exposure is associated with a second nitric oxide-independent but calcium-dependent early depressant mechanism that is manifested by reduced contractile and cAMP response to beta-adrenergic stimulation.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
L-アルギニンメチルエステル 二塩酸塩, ≥98.0% (AT)