コンテンツへスキップ
Merck
  • Intercalation and controlled release of pharmaceutically active compounds from a layered double hydroxide.
Intercalation and controlled release of pharmaceutically active compounds from a layered double hydroxide.
Chemical communications (Cambridge, England) (2002-09-21)
A I Khan, L Lei, A J Norquist, D O'Hare
要旨
A series of pharmaceutically active compounds including diclofenac, gemfibrozil, ibuprofen, naproxen, 2-propylpentanoic acid, 4-biphenylacetic acid and tolfenamic acid can be reversibly intercalated into a layered double hydroxide, initial studies suggest that these materials may have application as the basis of a novel tuneable drug delivery system.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
4-ビフェニル酢酸, 98%