コンテンツへスキップ
Merck
Total synthesis of a cyclic adenosine 5'-diphosphate ribose receptor agonist.
The Journal of organic chemistry (2012-01-31)
Joanna M Swarbrick, Barry V L Potter
要旨
Stable cyclic adenosine 5'-diphosphate ribose (cADPR) analogues are chemical biology tools that can probe the Ca(2+) release mechanism and structure-activity relationships of this emerging potent second messenger. However, analogues with an intact "northern" ribose have been inaccessible due to the difficulty of generating the sensitive N1-ribosyl link. We report the first total synthesis of the membrane permeant, hydrolytically stable, cADPR receptor agonist 8-Br-N1-cIDPR via regio- and stereoselective N1-ribosylation of protected 8-bromoinosine.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
環状アデノシン二リン酸リボース, ≥90% (HPLC), lyophilized powder
Sigma-Aldrich
ジ-tert-ブチルN,N-ジイソプロピルホスホロアミダイト, 95%