コンテンツへスキップ
Merck
  • Cytotoxic palladium(II) complexes of 8-aminoquinoline derivatives and the interaction with human serum albumin.
Cytotoxic palladium(II) complexes of 8-aminoquinoline derivatives and the interaction with human serum albumin.
Journal of inorganic biochemistry (2011-11-25)
Liangliang Yan, Xiaoyong Wang, Yanqing Wang, Yangmiao Zhang, Yizhi Li, Zijian Guo
要旨
Palladium(II) complexes are potential antitumor metallodrugs for their chemical resemblance to platinum(II) complexes. Two palladium(II) complexes (1 and 2) in the formula of [PdL(n)Cl] [L(1) = N-(tert-butoxycarbonyl)-l-methionine-N'-8-quinolylamide, L(2) = L-alanine-N'-8-quinolylamide] have been synthesized accordingly. The structures of the complexes were fully characterized by X-ray crystallography. The palladium(II) center in 1 is coordinated by two N atoms and an S atom from L(1) with one chloride anion as the leaving group; while that in 2 is coordinated by three N atoms from L(2) with one chloride anion as the leaving group. The interaction between complex 1 and human serum albumin (HSA) has been investigated using fluorescence and circular dichroism spectroscopies. The complex seems to react with HSA chiefly through hydrophobic and electrostatic interactions, and it does not alter the α-helical nature of HSA. The cytotoxicity of these complexes has been tested against the human cervical cancer (HeLa), human mammary cancer (MCF-7), and human lung cancer (A-549) cell lines. Complex 1 displays a cytotoxic activity comparable to that of cisplatin, but complex 2 is less active than cisplatin.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
8-アミノキノリン, 98%