コンテンツへスキップ
Merck
A unique enzyme from Saccharothrix sp. catalyzing D-amino acid transfer.
Biochimica et biophysica acta (1997-01-04)
Y Watanabe, T Muro, A Sugihara, Y Shimada, T Nagao, S Takenishi, Y Tominaga
要旨
A newly isolated actinomycete belonging to Saccharothrix sp. was found to produce a unique enzyme catalyzing D-amino acid transfer. The enzyme, which was tentatively named D-amino acid transferase, was purified 2600-fold to electrophoretic homogeneity and the molecular mass was 41 kDa. The enzyme was D-configuration specific and recognized aromatic D-amino acid esters to form oligo D-amino acid esters. D-Phenylalanine ester was favored as substrate over other D-amino acid esters. The optimum conditions for oligo D-phenylalanine ester formation by D-amino acid transferase were pH 7.0 and 40 degrees C. The enzyme was inhibited by DAN, EPNP and DFP.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
L-フェニルアラニンメチルエステル 塩酸塩, 98%
Sigma-Aldrich
D-フェニルアラニンメチルエステル 塩酸塩, 98%