コンテンツへスキップ
Merck
Dual effect of synthetic plasmin substrates on plasminogen activation.
Biochimica et biophysica acta (1995-03-15)
K Kolev, W G Owen, R Machovich
要旨
The effect of plasmin substrates D-valyl-L-leucyl-lysine-p-nitroanilide (S-2251) and H-D-norleucyl-hexahydrotyrosyl-lysine-p-nitro-anilide (Spectrozyme-PL) on the rate of activation of native human plasminogen in physiological salt solution is studied. Plasminogen activation by two-chain urokinase-type plasminogen activator (urokinase), two-chain tissue-type plasminogen activator (tc-tPA) or trypsin, but not by single chain tPA (sc-tPA) is increased 5- to 10-fold by both substrates, as determined by electrophoretic and spectrophotometric kinetic analysis. The amidolytic activity of sc-tPA, on the other hand, is inhibited by the plasmin substrates in a non-competitive manner (K1 of 6.4 . 10(-4) M for S-2251 and 2.9 . 10(-4) M for Spectrozyme-PL), whereas urokinase and tc-tPA activities are not affected. It is concluded that plasmin substrates containing a lysine residue have a general capacity to enhance plasminogen activation presumably by inducing a conformational change in the native zymogen in a manner similar to 6-aminohexanoate, while the same substrates are inhibitory both on the amidolytic activity of sc-tPA and the activation of native and des1-77-plasminogen by sc-tPA.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
D-Val-Leu-Lys 4-ニトロアニリド 二塩酸塩, plasmin substrate