コンテンツへスキップ
Merck
Gallocyanin-chrome alum counterstaining of Golgi-Kopsch impregnations.
Stain technology (1983-05-01)
S B Tieman
要旨
A simple technique is described for counterstaining Golgi-Kopsch impregnations. The sections are first stabilized by the method of Geisert and Updyke and then stained in 0.15% gallocyanin and 5% chromium potassium sulfate for 45 minutes at 55-60 C. The sections are then rinsed, dehydrated to 70% ethanol, cleared in terpineol, mounted and coverslipped. This procedure results in a light to medium blue stain of those cells not impregnated by the silver chromate. The major advantages of this procedure over earlier methods are: (1) the counterstain does not fade and (2) since no differentiation is required, many sections may be stained simultaneously.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
硫酸クロム(III)カリウム 十二水和物, ACS reagent, ≥98%
Sigma-Aldrich
硫酸クロム(III)カリウム 十二水和物, purum p.a., ≥98.5% (RT)
Sigma-Aldrich
Gallocyanine, Dye content 90 %
Sigma-Aldrich
硫酸クロム(III)カリウム 十二水和物, SAJ first grade, ≥97.0%