- Trapezium resection with mersilene suspension sling.
Trapezium resection with mersilene suspension sling.
Hand clinics (2012-11-22)
Marwan A Wehbé, Rita M Ciccarello, Jennifer L Reitz
PMID23168026
要旨
The reason there are numerous techniques for thumb carpometacarpal arthroplasty is that none of them are perfect. Sometimes the simplest procedures work best. This article presents a simple alternative, using a readily available suture to stabilize the thumb after resection of the trapezium, with long-term success.