コンテンツへスキップ
Merck
  • Marked resistance of femoral adipose tissue blood flow and lipolysis to adrenaline in vivo.
Marked resistance of femoral adipose tissue blood flow and lipolysis to adrenaline in vivo.
Diabetologia (2012-08-18)
K N Manolopoulos, F Karpe, K N Frayn
要旨
Fatty acid entrapment in femoral adipose tissue has been proposed to prevent ectopic fat deposition and visceral fat accumulation, resulting in protection from insulin resistance. Our objective was to test the hypothesis of femoral, compared with abdominal, adipose tissue resistance to adrenergic stimulation in vivo as a possible mechanism. Regional fatty acid trafficking, along with the measurement of adipose tissue blood flow (ATBF) with (133)Xe washout, was studied with the arteriovenous difference technique and stable isotope tracers in healthy volunteers. Adrenergic agonists (isoprenaline, adrenaline [epinephrine]) were infused either locally by microinfusion or systemically. Local microinfusion of adrenoceptor antagonists (propranolol, phentolamine) was used to characterise specific adrenoceptor subtype effects in vivo. Femoral adipose tissue NEFA release and ATBF were lower during adrenaline stimulation than in abdominal tissue (p < 0.001). Mechanistically, femoral adipose tissue displayed a dominant α-adrenergic response during adrenaline stimulation. The α-adrenoceptor blocker, phentolamine, resulted in the 'disinhibition' of the femoral ATBF response to adrenaline (p < 0.001). Fatty acids, once stored in femoral adipose tissue, are not readily released upon adrenergic stimulation. Femoral adipose tissue resistance to adrenaline may contribute to the prevention of ectopic fatty acid deposition.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
フェントラミン 塩酸塩, ≥98% (TLC), powder
Sigma-Aldrich
フェントラミン メタンスルホン酸塩, ≥98% (TLC), powder