コンテンツへスキップ
Merck
Fully automated production of 11C-doxepin for PET imaging histamine H1 receptor.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2012-01-10)
Hancheng Cai, Thomas J Mangner, Otto Muzik, Xin Lu, Pulak K Chakraborty, Diane C Chugani, Harry T Chugani
要旨
(11)C-Doxepin is an established positron emission tomography (PET) probe for imaging the histamine H1 receptor, which is associated with various neurological disorders and allergic diseases. A fully automated current Good Manufacturing Practices (cGMP)-compliant radiosynthesis is therefore desirable in order to facilitate clinical PET studies. We report here a fully automated production method for (11)C-doxepin using a multipurpose PET module for clinical use. (11)C-Doxepin was radiosynthesized by N-[(11)C]methylation of nordoxepin using [(11)C]methyl iodide in DMF solvent, and then purified by HPLC, and finally reformulated with solid phase extraction (SPE) using a cGMP-compliant automated multipurpose PET module developed in house. The final product was analyzed and subjected to quality control according to current US Pharmacopeia requirements. The radiochemical yield (decay corrected) of (11)C-doxepin for clinical use was 47.0 ± 5.2% (n = 12) based on [(11)C]methyl iodide, moreover the radiochemical purity of (11)C-doxepin was more than 97.5% with 1,200 ± 500 Ci/mmol specific activity(end of production). The total production time of (11)C-doxepin was 37 min from end of bombardment (EOB) with the final product passing all tests under cGMP requirements for clinical use. A simplified and reliable fully automated production of (11) C-doxepin for clinical use was developed, allowing the synthesis of the tracer with high yield using a cGMP-compliant module and procedure. The success of this approach could make the PET tracer (11) C-doxepin more accessible for clinical studies.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ドキセピン 塩酸塩, ~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder
Supelco
Doxepin hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
システム適合性用ドクサピン, European Pharmacopoeia (EP) Reference Standard