コンテンツへスキップ
Merck
  • Olopatadine 0.6% nasal spray protects from vasomotor challenge in patients with severe vasomotor rhinitis.
Olopatadine 0.6% nasal spray protects from vasomotor challenge in patients with severe vasomotor rhinitis.
American journal of rhinology & allergy (2011-08-09)
Peter K Smith, Joel Collins
要旨
Vasomotor rhinitis (VMR) is a hypersensitivity syndrome with heightened reactivity to environmental triggers. Twenty-two patients with severe VMR were treated nasally with either normal saline or 0.6% olopatadine and challenged nasally with a hyperosmolar mannitol solution. Treatment with 0.6% olopatadine resulted in an improvement in instantaneous nasal symptom scores at 5 and 30 minutes (p < 0.01) compared with baseline and at 30 minutes after hyperosmolar challenge compared with saline-pretreated individuals (p < 0.01). There was also an improvement in nasal peak inspiratory flow rate at 30 minutes after hyperosmolar challenge compared with saline-pretreated individuals (p < 0.01). In this patient population 0.6% olopatadine appears to be efficacious in symptom reduction in VMR and protects from hyperosmolar challenge.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
オロパタジン 塩酸塩, ≥98% (HPLC)