コンテンツへスキップ
Merck
Mild addition of nucleophiles to pyridine-N-oxides.
Organic letters (2011-03-08)
Allyn T Londregan, Sandra Jennings, Liuqing Wei
要旨
A general and facile one-pot procedure for the synthesis of 2-substituted pyridines from the corresponding pyridine-N-oxides and nucleophiles is presented as a mild alternative to S(N)Ar chemistry. A variety of nucleophiles and heterocyclic-N-oxides participate effectively in this transformation, which uses the phosphonium salt, PyBroP, as a means of substrate activation.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ピリジンN-オキシド, 95%