コンテンツへスキップ
Merck
  • Enhanced formation of methane in plant cell cultures by inhibition of cytochrome c oxidase.
Enhanced formation of methane in plant cell cultures by inhibition of cytochrome c oxidase.
Plant, cell & environment (2010-11-11)
Asher Wishkerman, Steffen Greiner, Miklós Ghyczy, Mihály Boros, Thomas Rausch, Katharina Lenhart, Frank Keppler
要旨
The claim of methane (CH₄) formation in plants has caused much controversy and debate within the scientific community over the past 4 years. Here, using both stable isotope and concentration measurements, we demonstrate that CH₄ formation occurs in plant cell cultures that were grown in the dark under sterile conditions. Under non-stress conditions the plant cell cultures produced trace amounts [0.3-0.6 ng g⁻¹ dry weight (DW) h⁻¹] of CH₄ but these could be increased by one to two orders of magnitude (up to 12 ng g⁻¹ DW h⁻¹) when sodium azide, a compound known to disrupt electron transport flow at the cytochrome c oxidase (complex IV) in plant mitochondria, was added to the cell cultures. The addition of other electron transport chain (ETC) inhibitors did not result in significant CH₄ formation indicating that a site-specific disturbance of the ETC at complex IV causes CH₄ formation in plant cells. Our study is an important first step in providing more information on non-microbial CH₄ formation from living plants particularly under abiotic stress conditions that might affect the electron transport flow at the cytochrome c oxidase in plant mitochondria.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
サリチルヒドロキサム酸, 99%