コンテンツへスキップ
Merck
  • Protective effects of platinum nanoparticles against UV-light-induced epidermal inflammation.
Protective effects of platinum nanoparticles against UV-light-induced epidermal inflammation.
Experimental dermatology (2010-09-04)
Yoko Yoshihisa, Ayumi Honda, Qing-Li Zhao, Teruhiko Makino, Riichiro Abe, Kotaro Matsui, Hiroshi Shimizu, Yusei Miyamoto, Takashi Kondo, Tadamichi Shimizu
要旨
Intracellular reactive oxygen species (ROS) and apoptosis play important roles in the ultraviolet (UV)-induced inflammatory responses in the skin. Metal nanoparticles have been developed to increase the catalytic activity of metals, which is because of the large surface area of smaller particles. Platinum nanoparticles (nano-Pt) protected by poly acrylic acid were manufactured by reduction with ethanol. A marked increase in ROS production was observed in UV-treated HaCaT keratinocytes cell lines, while a decrease in ROS production was observed in nano-Pt-treated cells. Pretreatment of the cells with nano-Pt also caused a significant inhibition of UVB- and UVC-induced apoptosis. Furthermore, we found that mice treated with nano-Pt gel prior to UV irradiation showed significant inhibition of UVB-induced inflammation and UVA-induced photoallergy compared to UV-irradiated control mice. These results suggest that nano-Pt effectively protects against UV-induced inflammation by decreasing ROS production and inhibiting apoptosis in keratinocytes.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ロメフロキサシン 塩酸塩