コンテンツへスキップ
Merck
  • Syntheses of amorphous and crystalline cupric sulfide nanoparticles and study on the specific activities on different cells.
Syntheses of amorphous and crystalline cupric sulfide nanoparticles and study on the specific activities on different cells.
Chemical communications (Cambridge, England) (2010-04-09)
Yuming Guo, Jie Zhang, Lin Yang, Huajie Wang, Feifei Wang, Zhi Zheng
要旨
Copper sulfide amorphous nanoparticles and nanocrystals were prepared successfully by a special process. These CuS nanoparticles could specifically and significantly induce the apoptosis and inhibit the proliferation of human cancer cells rather than normal cells. Moreover, the biological activities of these nanoparticles are related to their polymorphs.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
硫化銅(I), powder, −325 mesh
Sigma-Aldrich
硫化銅(I), anhydrous, powder, 99.99% trace metals basis