コンテンツへスキップ
Merck
  • Inhibition of specific electron transport pathways leads to oxidative stress and decreased Candida albicans proliferation.
Inhibition of specific electron transport pathways leads to oxidative stress and decreased Candida albicans proliferation.
Journal of bioenergetics and biomembranes (2006-10-21)
Fernando Ruy, Anibal E Vercesi, Alicia J Kowaltowski
要旨
Candida parapsilosis mitochondria contain three respiratory chains: the classical respiratory chain (CRC), a secondary parallel chain (PAR) and an "alternative" oxidative pathway (AOX). We report here the existence of similar pathways in C. albicans. To observe the capacity of each pathway to sustain yeast growth, C. albicans cells were cultured in the presence of inhibitors of these pathways. Antimycin A and KCN totally abrogated yeast growth, while rotenone did not prevent proliferation. Furthermore, rotenone promoted only partial respiratory inhibition. Lower concentrations of KCN that promote partial inhibition of respiration did not inhibit yeast growth, while partial inhibition of respiration with antimycin A did. Similarly, AOX inhibitor BHAM decreased O(2) consumption slightly but completely stunted cell growth. Reactive oxygen species production and oxidized glutathione levels were enhanced in cells treated with antimycin A or BHAM, but not rotenone or KCN. These findings suggest that oxidative stress prevents C. albicans growth.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ベンズヒドロキサム酸, 99%