コンテンツへスキップ
Merck
  • Chemical composition and biological activity of the volatiles of Anthemis melampodina and Pluchea dioscoridis.
Chemical composition and biological activity of the volatiles of Anthemis melampodina and Pluchea dioscoridis.
Phytotherapy research : PTR (2002-04-05)
Mary H Grace
要旨
The volatile fractions obtained by hydrodistillation of the fresh leaves of Anthemis melampodina and Pluchea dioscoridis were analysed by GC-MS technique. Of 38 components identified in the volatile oil of A. melampodina, santolinatriene was the major component (27.33%). The oil was characterized by a high percentage of monoterpene hydrocarbons (49.94%) while sesquiterpene hydrocarbons and oxygenated sesquiterpenes represented only 7.41% and 11.43% of the oil. 36 components were identified in the volatile oil of P. dioscoridis. Farnesol was the major component (16.50%) accompanied by a high percentage of sesquiterpene alcohols. Oxygenated sesquiterpenes (26.43%) and sesquiterpene hydrocarbons (39.43%) represented the main constituents in the oil. P. dioscoridis showed a marked mosquito larvicidal activity against Culex pipiens (LC(50) 71.86 ppm), while A. melampodina was moderately active (LC(50) 139.42 ppm).
材料
製品番号
ブランド
製品内容
Supelco
シフェノトリン, PESTANAL®, analytical standard, sum of isomers