コンテンツへスキップ
Merck
  • Increased survival of dopaminergic neurons by rasagiline, a monoamine oxidase B inhibitor.
Increased survival of dopaminergic neurons by rasagiline, a monoamine oxidase B inhibitor.
Neuroreport (1998-04-29)
J P Finberg, T Takeshima, J M Johnston, J W Commissiong
要旨
Both deprenyl and rasagiline (R(+)-N-propargyl-1-aminoindane mesylate), at a concentration of 1-10 microM, increased survival in vitro of rat E14 mesencephalic dopaminergic neurons that had been primed with 10% serum for 12 h (p < 0.05). Rasagiline, but not deprenyl, also increased total neuronal (MAP2-positive) survival (p < 0.05) Under serum-free conditions, rasagiline, but not deprenyl, retained its neuroprotective action on dopaminergic neurones. GABAergic neurons were not affected by either deprenyl or rasagiline. Clorgyline, an MAO-A inhibitor, did not exert any of these effects. The protective action of rasagiline on dopaminergic neurons, even under stringent serum-free conditions, is striking, and warrants further investigation for a role in the treatment of Parkinson's disease.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ラサギリン メシル酸塩, ≥98% (HPLC)