コンテンツへスキップ
Merck
  • Green fluorescent protein emission obscures metabolic fluorescent lifetime imaging of NAD(P)H.
Green fluorescent protein emission obscures metabolic fluorescent lifetime imaging of NAD(P)H.
Biomedical optics express (2019-10-01)
Elisa M York, Nicholas L Weilinger, Jeffrey M LeDue, Brian A MacVicar
要旨
Autofluorescence of endogenous molecules can provide valuable insights in both basic research and clinical applications. One such technique is fluorescence lifetime imaging (FLIM) of NAD(P)H, which serves as a correlate of glycolysis and electron transport chain rates in metabolically active tissue. A powerful advantage of NAD(P)H-FLIM is the ability to measure cell-specific metabolism within heterogeneous tissues. Cell-type specific identification is most commonly achieved with directed green fluorescent protein (GFP) expression. However, we demonstrate that NAD(P)H-FLIM should not be analyzed in GFP-expressing cells, as GFP molecules themselves emit photons in the blue spectrum with short fluorescence lifetimes when imaged using two-photon excitation at 750 nm. This is substantially different from the reported GFP emission wavelength and lifetime after two-photon excitation at 910 nm. These blue GFP photons are indistinguishable from free NAD(P)H by both emission spectra and fluorescence lifetime. Therefore, NAD(P)H-FLIM in GFP-expressing cells will lead to incorrect interpretations of metabolic rates, and thus, these techniques should not be combined.
材料
製品番号
ブランド
製品内容
Millipore
緑色蛍光タンパク質