コンテンツへスキップ
Merck
  • ER-resident oxidoreductases are glycosylated and trafficked to the cell surface to promote matrix degradation by tumour cells.
ER-resident oxidoreductases are glycosylated and trafficked to the cell surface to promote matrix degradation by tumour cells.
Nature cell biology (2020-10-21)
Manon Ros, Anh Tuan Nguyen, Joanne Chia, Son Le Tran, Xavier Le Guezennec, Ruth McDowall, Sergey Vakhrushev, Henrik Clausen, Martin James Humphries, Frederic Saltel, Frederic André Bard
要旨
Tumour growth and invasiveness require extracellular matrix (ECM) degradation and are stimulated by the GALA pathway, which induces protein O-glycosylation in the endoplasmic reticulum (ER). ECM degradation requires metalloproteases, but whether other enzymes are required is unclear. Here, we show that GALA induces the glycosylation of the ER-resident calnexin (Cnx) in breast and liver cancer. Glycosylated Cnx and its partner ERp57 are trafficked to invadosomes, which are sites of ECM degradation. We find that disulfide bridges are abundant in connective and liver ECM. Cell surface Cnx-ERp57 complexes reduce these extracellular disulfide bonds and are essential for ECM degradation. In vivo, liver cancer cells but not hepatocytes display cell surface Cnx. Liver tumour growth and lung metastasis of breast and liver cancer cells are inhibited by anti-Cnx antibodies. These findings uncover a moonlighting function of Cnx-ERp57 at the cell surface that is essential for ECM breakdown and tumour development.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ゼラチン 溶液, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
GPX1 human, recombinant, expressed in E. coli, His tagged, >90% (SDS-PAGE)