コンテンツへスキップ
Merck
  • An optimized mixed-mode stationary phase based on silica monolith particles for the separation of peptides and proteins in high-performance liquid chromatography.
An optimized mixed-mode stationary phase based on silica monolith particles for the separation of peptides and proteins in high-performance liquid chromatography.
Journal of separation science (2019-10-07)
Genlin Sun, Ashraf Ali, Yune Sung Kim, Jeong Soo Kim, Hyun Joo An, Won Jo Cheong
要旨
A phase with both hydrophobic and hydrophilic functionalities has been synthesized by modification of ground silica monolith particles with C18 and 1-[3-(trimethoxysilyl)propyl] urea ligands. A series of phases was prepared by changing the ratio of the two ligands to determine the optimal ratio in view of separation efficiency. The resultant optimized stationary phase was packed in narrow-bore glass-lined stainless-steel columns (1 × 300 mm and 2.1 × 100 mm) and used for the separation of synthetic peptides and proteins. The average numbers of theoretical plates (N) of 52 100/column (174 000/m, 5.75 µm plate height) and 35 500/column (118 000/m, 8.47 µm plate height) were achieved with the 300 mm column at a flow rate of 25 µL/min (0.86 mm/s) in 60:40 v/v acetonitrile/30 mM aqueous ammonium formate for the mixture of peptides (Thr-Tyr-Ser, Val-Ala-Pro-Gly, angiotensin I, isotocin, and bradykinin) and for the mixture of proteins (myoglobin, human serum albumin, and insulin), respectively. Fast analysis of the peptides and proteins was also carried out at a flow rate of 0.9 mL/min (6.88 mm/s) with the 100 mm column and all the analytes were eluted within 2 min with good separation efficiency.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
トリメトキシ(オクタデシル)シラン, technical grade
Sigma-Aldrich
1-[3-(トリメトキシシリル)プロピル]尿素, 97%