コンテンツへスキップ
Merck
  • Polyamines differentially inhibit cyclic AMP-dependent protein kinase-mediated phosphorylation in the brain of the tobacco hornworm, Manduca sexta.
Polyamines differentially inhibit cyclic AMP-dependent protein kinase-mediated phosphorylation in the brain of the tobacco hornworm, Manduca sexta.
Journal of neurochemistry (1988-11-01)
W L Combest, T J Bloom, L I Gilbert
要旨
The effects of the naturally occurring polyamines spermine and spermidine on phosphorylation promoted by cyclic AMP (cAMP)-dependent protein kinase (PK) (cAMP-PK; EC 2.7.1.37) were studied using the brain of the tobacco hornworm, Manduca sexta. Four particulate-associated peptides (280, 34, 21, and 19 kilodaltons) in day 1 pupal brains are endogenous substrates for a particulate type II cAMP-PK. These phosphoproteins are present in brain synaptosomal, as well as microsomal, particulate fractions but are not present in the cytosol. They are distributed throughout the CNS and PNS and are present in several nonneuronal tissues as well. Phosphorylation of these proteins via cAMP-PK was inhibited markedly by micromolar concentrations of spermine and spermidine. Other particulate-associated peptides phosphorylated via a Ca2+/calmodulin-PK or Ca2+ and cAMP-independent PKs were unaffected by polyamines, whereas the phosphorylation of a 260-kilodalton peptide was markedly enhanced. Spermine did not exert its inhibitory effect indirectly by enhancement of cAMP or ATP hydrolysis or via proteolysis, but its action appears to involve a substrate-directed inhibition of cAMP-PK-promoted phosphorylation as well as enhanced dephosphorylation. Although addition of spermine resulted in marked ribosome aggregation in synaptosomal and microsomal particulate fractions, this phenomenon was not involved in the inhibition of cAMP-PK-promoted phosphorylation.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
Hexafluoropropene, trimer