Merck
HomeBiopharmaceutical CharacterizationProtein Fingerprinting of a Viral Vector, AAV5
Protein Fingerprinting of a Viral Vector, AAV5
Geoffrey Rule, Kevin Ray, Uma Sreenivasan, Cory Muraco, Pei Liu
Merck
Article from Analytix Reporter - Issue 16
Introduction
The culture and use of adeno-associated virus (AAV) as a gene delivery device has seen much interest in recent years as a strategy for delivering targeted gene therapies towards muscle, nerve, liver, and eye disorders. This includes different AAV serotypes that target specific tissues as well as genetically engineered hybrid types with altered tissue specificity.
In-depth characterization of the viral capsid proteins, as well as the genomic content, is essential to verify critical quality attributes of these particles. Both the amino acid sequence as well as post-translational modifications (PTMs) that are found on the viral capsid proteins can have an impact on the tissue tropism, efficacy, and immunogenicity of AAV.1 PTMs that have been identified on AAV include phosphorylation, SUMOylation, ubiquitination, acetylation, methylation, and glycosylation.2
The USP released draft guidelines3 for the analytical characterization of viral vectors in 2022 to provide method starting points for determination of critical quality attributes. A variety of tests are described to characterize the identity, purity, concentration, and potency of these viral vectors, among other traits. For determination of capsid identity, starting methods are provided that use Western blotting, reversed-phase HPLC with UV detection, and HPLC with MS detection. The latter approach describes the use of both intact mass analysis of the capsid proteins, as well as amino acid sequence analysis using peptide mapping. Here we describe our work to develop methods for protein fingerprinting of AAV serotype 5 using both intact mass analysis and peptide mapping. Several post- translational modifications of the viral proteins VP1, VP2 and VP3 were identified.
Experimental Methods
A system suitability mix, MSRT1, was prepared according to the instructions on the data sheet but with a final acetonitrile concentration of 1.6% (v/v). The injection volume was 10 µL. This solution is a mix of 14 isotopically labelled peptides injected prior to injection of samples to verify instrument performance.
AAV Production
AAV5 subtype (Q9YIJ1) was produced in HEK293 non-T VirusExpress® 293 AAV Production Cells using EX-CELL® CD HEK293 Viral Vector Medium. Three transfection plasmids carrying: 1. Replication and capsid genes; 2. certain adenovirus genes on a helper plasmid and 3. a plasmid with the gene of interest, in this case green fluorescence protein (GFP), were used. After three days of transfection, full AAV capsids, as well as product related impurities such as empty capsids and capsids filled with host DNA fragments or plasmid backbones, were produced. The HEK cells were then lysed with detergent, treated with benzonase/magnesium, to digest all unused plasmid DNA and host nucleic acids, and finally treated with 0.5 M sodium chloride. The clarified lysate was affinity purified on a fast protein liquid chromatography (FPLC) system (ÄKTA) using commercially available resin, from host cell proteins and digested nucleic acids. This process resulted in viral particle (vp) concentrations between 4E+12 and 2E+13 vp/mL as determined by droplet digital polymerase chain reaction (ddPCR).
Intact Mass Analysis
AAV particles were treated with 10% acetic acid for 15 minutes to dissociate AAV capsid assembly before centrifugation at 12,000 rpm for five minutes. Approximately 0.6 µg of total AAV capsid protein was injected for each run.
Chromatography was performed on BIOshell™ A400 Protein C4, 3.4 µm particle, and BIOshell™ IgG 1000 Å C4, 2.7 µm particle columns, along with a competitor column, mentioned in the draft guideline, for comparison. All had dimensions of 100 x 2.1 mm. Mass spectrometry was conducted on a Waters™ Xevo™ G2-S QTof in positive ion mode. The chromatographic conditions and instrument parameters used for the applied LC-UV-MS method are shown in Tables 1 and 2 below.
LC Conditions
Instrument:ACQUITY™ Premier UPLC
Columns:BIOshell™ A400 Protein C4, 100 x 2.1, 3.4 µm (66825-U); BIOshell™ IgG 1000 Å C4, 100 x 2.1, 2.7 µm (63288-U)
Mobile phase:[A] Water containing 5% (v/v) acetonitrile with 0.1% TFA; [B] Acetonitrile (0.1% v/v TFA)
Gradient:Time (min)A%B% 
 0.08020 
 1.06832 
 16.06436 
 20.02080 
 21.52080 
 22.08020 
 30.08020 
Flow rate:0.2 mL/min
Column temp.:80 °C
Detector:UV @ 280 nm and mass spectrometry (Table 2)
Injection:5 µL
Sample:As described in text
Table 1.HPLC conditions for intact mass analysis
Mass Spectrometer Conditions
Instrument:Waters™ Xevo™ G2-S QTof
Polarity:Positive ion
Capillary (kV)3.0 kV
Sampling Cone120 V
Source Offset120 V
Source Temperatures125 °C
m/z range:500-4000
Table 2.MS Conditions used for intact mass analysis
Peptide Mapping
Digestion of purified AAV particles was conducted with both trypsin and, separately, chymotrypsin, using a Low-Artifact Digestion Buffer and a filter-assisted, sample preparation protocol. For details on this protocol, please see the online application note.4
BIOshell™ A160 Peptide C18, 2.7 µm particle column and an identical BIOshell™ column with 2 µm particles were used with gradient conditions outlined in the USP draft guidelines.3 Both columns were 150 x 2.1 mm in dimension. For comparison, a competitor C18 column, mentioned in the draft guidelines, was evaluated using the same conditions (Table 3) and column dimensions.
Mass spectrometry was conducted on a Thermo QE Plus in positive ion mode using a scan range of 200 to 2000 m/z and data dependent MS2 of the top ten ions (Table 4).
LC Conditions 
Instrument:ACQUITY™ UPLC
Columns:BIOshell™ A160 Peptide C18, 2.7 µm (66905-U); BIOshell™ A160 Peptide C18, 2 µm (67243-U); Competitor C18 130 Å, 2.5 µm FPP column All 150 x 2.1 mm
Mobile phase:[A] 0.1% Formic acid in water (v/v); [B] 0.1% Formic acid in acetonitrile (v/v)
Gradient:Time (min)A%B% 
 0.0973 
 0.5973 
 50.04555 
 50.11090 
 55.01090 
 55.1973 
 75.0973 
Flow rate:0.25 mL/min
Column temp.:40 °C
Pressure:2890 psi (2.7 µm column); 5725 psi (2 µm column); 3555 psi (2.5 µm column) at start of run.
Detector:MS (Table 4)
Injection:5 µL
Sample(s):As described in text
Table 3.HPLC conditions used for peptide mapping
MS Conditions
Instrument:Thermo QE Plus
Polarity:Positive ion
Spray Voltage:3.5 kV
Capillary Temperature: 320 °C
Sheath Gas:10
Aux gas:5
S-Lens50 V
m/z range:200-2000
ddMS2:Top 10
Table 4.Mass spectrometer conditions used for peptide mapping
Results
Protein Fingerprinting
The draft USP guidelines provided a starting point for MS characterization of viral vectors by both intact capsid fingerprinting and peptide mapping. The draft guideline also describes an additional method, using UV detection and a 2-hour chromatographic run, for determination of capsid stoichiometry. While we did not replicate this method, we did use UV detection in conjunction with mass spectrometry fingerprinting of the intact viral capsids over a shorter 30-minute run. Integration of the UV detected peaks was then used to evaluate capsid stoichiometry. The combined LC- UV-MS analysis provided a convenient, one-method assessment of fingerprint and stoichiometry in a shorter run time.
Intact Mass Analysis of VP1, VP2, and VP3
The comparison of columns for separation of the intact capsid proteins is shown in Figure 1. Both the BIOshell™ A400 C4 and BIOshell™ IgG 1000 Å C4 columns provided good retention of the proteins using the gradient described in the draft guideline. Separation of VP1 from VP2 and VP3 was also achieved. The BIOshell™ A400 C4 column also provided partial separation of a VP3-clip from VP3. The competitor column gave faster elution with the same conditions but did not show a distinct peak for VP1. Deconvolution of the peaks observed on the BIOshell™ A400 C4 column resulted in the mass determinations as shown in Figure 2.
Both VP1 and VP3 were observed to be highly acetylated while VP2 showed a degree of phosphorylation. The presence of 0.1% TFA in the mobile phases was found to result in a substantial amount of TFA adduction on each of the proteins. Interestingly, other AAV serotypes (AAV2, AAV8) we have examined have not shown this same degree of TFA adduction. Replacing the TFA with 0.1% formic acid resulted in a loss of chromatographic separation of the three capsid proteins although the TFA adduction was eliminated (data not shown). The use of difluoroacetic acid (DFA) could be evaluated as a compromise between chromatographic separation and MS sensitivity.
A comparison of total ion current (TIC) profiles of intact AAV5 capsid protein to assess its retention and separation on three different columns. Both the BIOshell™ A400 C4 and BIOshell™ IgG 1000 Å C4 columns provided good retention of the proteins using the gradient described in the draft guideline. Separation of VP1 from VP2 and VP3 was also achieved
Figure 1.Total ion current (TIC) profile comparison of AAV5 capsid protein retention and separation on three columns evaluated.
Deconvoluted spectra of each of the four dominant peaks observed during the intact capsid protein analysis of AAV5 on the BIOshell™ A400 Protein C4 column resulted in the mass determinations as shown. Peak 1 at 10.06 min major mass traces are VP2 65282, VP2+phos 62362, and VP2+TFA 65396; Peak 2 at 10.64 min major mass traces are VP3+Ac 59462 and VP3+Ac+TFA 59576, Peak 3 at approx. 11 min major mass traces are VP3 clip+Ac 56123, VP3 clip+Ac+TFA 56237; Peak 4 at 12.96 min major mass traces areVP1+Ac 80335 and VP1+Ac+TFA 80447.
Figure 2.Deconvoluted spectra from each of four dominant peaks observed on the BIOshell™ A400 Protein C4 column.
A clipped form of VP3 was also observed, corresponding to the cleavage of the aspartic acid (695) - proline (696). This bond is known to be particularly labile to hydrolysis under acidic conditions and elevated temperature, and similar clip proteoforms have been reported by others.5,6 Table 5 shows the close agreement of the observed masses and the theoretical masses for the capsid proteins.
Viral ProteinAA seqModificationTheoretical mass (Da)Observed mass (Da)Mass error
VP12-724N-term Acetylation80336803350.0012%
VP2138-724 65283652820.0015%
VP3194-724N-term Acetylation59463594620.0017%
VP3-clip194-695N-term Acetylation56125561230.0036%
Table 5.Observed masses and the theoretical masses for the capsid proteins
Using the same conditions described above, but with UV detection, a measurement of the relative abundance of the four main peaks was made, as shown in Figure 3.
While the stoichiometry of AAV capsids is sometimes suggested to have a specific ratio of capsid proteins, such as 1:1:10, Wörner et al.7 nicely describe the process of capsid formation as being a stochastic sampling of capsid proteins available in the cell pool. AAV capsids are then made up of a highly heterogenous composition of three capsid proteins so that measured stoichiometry represents only an average composition. Combining both the UV and MS detection in the same run is an added convenience in the evaluation of samples.
Absorbance versus time plot showing the peaks for VP1, VP2, VP3, and VP3-clip obtained during the intact AAV5 capsid protein analysis using BIOshell™ A400 Protein C4 column and UV detection at 280 nm.
ProteinPeak AreaArea % (UV)Average count/VP
VP13033.62
VP2206524.315
VP3582168.443
VP3-clip3243.8
Figure 3.Stoichiometry determination using UV (280 nm) detection on the BIOshell™ A400 Protein C4 column.
Peptide Mapping
For the comparison here, we used the column and conditions suggested in the draft USP guidelines; a competitor column and gradient conditions shown in Tables 3 and 4, above. This gradient is delivered over a 50-minute period. On both the competitor and BIOshell™ columns, good sequence coverage was obtained using tryptic digestion but with slightly better sequence coverage on the BIOshell™ columns due to retention of several small, early eluting peptides including VVTK, ADEVAR, GEPVNR, SLRVK, RIDDHFPKR (Figure 4). Importantly, the N-terminal sequence of VP2, (APTGK) was also identified on these columns, in addition to the N-terminal sequences of VP1 and VP3 as shown in Figure 5.
A Venn diagram showing the number of peptides identified using each of the three columns evaluated- BIOshell™ A400 Protein C4, 3.4 µm particle, and BIOshell™ IgG 1000 Å C4, 2.7 µm particle columns, and a competitor C18 column.
A
 Sequence∆ppmRT (min)MS AreaProteinstartend
Polar/Early ElutersVVTK-1.841.894.24E+06VP1,2,3229232
ADEVAR-0.653.053.35E+05VP16772
GEPVNR-0.453.222.44E+05VP16166
SLRVK-1.074.111.88E+05VP1,2,3301305
RIDDHFPKR-0.777.981.99E+05VP1,2,3143151
VP2 N-terminusAPTGK1.181.588.11E+04VP2138142
B
Sequence∆ppmRTMS AreaProteinstartend
SGSVDGSNANAYFGYSTPWGYFDFNRFHSHWSPR0284.46E+06VP1,2,3252285
C
Sequence∆ppmRTMS AreaProteinstartend
STRTWVLPSYNNHQYREIK0.3615.31.02E+05VP1,2,3233251
STRTWVLPSYNNHQYR0.3915.51.03E+05VP1,2,3233248
DVYLQGPIWAKIPETGAHFHPSPAMGGFGLK0.9424.96.95E+04VP1,2,3600630
Figure 4.Venn diagram showing the number of peptides identified using each of the three columns evaluated
Four product ion spectra providing the sequence coverage of VP1, VP2, and VP3 N-termini as well as C-terminus of each indicating the observed mass fragments.
Four product ion spectra providing the sequence coverage of VP1, VP2, and VP3 N-termini as well as C-terminus of each indicating the observed mass fragments.
Four product ion spectra providing the sequence coverage of VP1, VP2, and VP3 N-termini as well as C-terminus of each indicating the observed mass fragments.
Four product ion spectra providing the sequence coverage of VP1, VP2, and VP3 N-termini as well as C-terminus of each indicating the observed mass fragments.
Figure 5.Product ion spectra providing sequence coverage of the N-termini of VP1 (A), VP2 (B), and VP3 (C) as well as C-terminus of each (D).
One large section of 60 amino acids was not covered by tryptic digestion alone, and so a separate digestion was performed with chymotrypsin to increase the overall sequence coverage. Similar multi-enzyme digestion of AAV capsids has been shown useful by others.8 Using the two enzymes, separately, increased the coverage to 100% on the BIOshell™ columns.
A summary of the PTMs identified, along with their percent abundance on each of the three columns, is shown in Table 6.
Both VP1 and VP3 were found to be 100% acetylated while at least four sites of phosphorylation were identified. Acetylation of the N-terminus of AAV capsids appears to be highly conserved across serotypes.2 In addition to the deamination site, four sites of oxidation were observed with all being less than 1% abundant than the unoxidized forms. Overall, the agreement in the abundances of the PTMs between the columns was very good.
ProteinResidue #ModificationPeptide Sequence% Abundance Competitor C18% Abundance BIOShell™ 2 µm% Abundance BIOShell™ 2.7 µm
VP11S1+AcetylationSFVDHPPDWLEEVGEGLR100100100
VP31S1+AcetylationSAGGGGPLGDNNQGADGVGNASGDWHCDSTWMGDR100100100
VP155N55+DeamidationGLVLPGYNYLGPGNGLDR28.230.225.2
VP1,2,3468M468+OxidationNWFPGPMGR0.530.560.67
VP1,2,3474W474+OxidationTQGWNLGSGVNR0.210.210.20
VP1,2,3568M568+OxidationVAYNVGGQMATNNQSSTTAPATGTYNLQEIVPGSVWMER0.730.760.59
VP1,2,3623M623+OxidationIPETGAHFHPSPAMGGFGLKHPPPMMLIK; IPETGAHFHPSPAMGGFGLK0.281.240.34
VP1,2,3530S530+PhosphorylationIFNSQPANPGTTATY5.705.805.90
VP1,2,3~648~S648+PhosphorylationMLIKNTPVPGNITSF; IKNTPVPGNITSF2.122.702.54
VP21-41PhosphorylationAPTGKRIDDHFPKRKKARTEEDSKPSTSSDAEAGPSGSQQL9.555.509.51
VP1,2155-227PhosphorylationTEEDSKPSTSSDAEAGPSGSQQLQIPAQPASSLGADTMSAGGGGPLGDNNQGADGVGNASGDWHCDSTWMGDR7.6811.599.10
Table 6.Post-translational modifications and their abundance determined
Conclusions
Several column comparisons were shown to demonstrate uses of the BIOshell™ line of columns for characterizing viral vectors, in this case AAV serotype 5. Conditions outlined in the USP draft guideline were used for both intact mass fingerprinting of viral capsids and for peptide mapping experiments, but we suggest that further improvements in chromatography might be made with additional gradient optimization.
The BIOshell™ columns, particularly the A400 Protein C4 column, have proven to be effective in separating the three capsid proteins of AAV5 for intact mass analysis and stoichiometry evaluation. In addition, the BIOshell™ A400 Protein C4 column provides partial separation of the VP3- clip from VP3. The competitor column also shows partial separation of VP3-clip from VP3 but with coelution of VP1.
The BIOshell™ A160 Peptide C18 columns, in both the 2.7 and 2.0 µm particle sizes, proved to be useful in retaining short, polar peptides to provide slightly improved sequence coverage over the competitor column. Retention of the N-terminus of VP2 was only provided by the BIOshell™ columns.
The USP draft conditions for the mobile phase and gradient conditions were used in both approaches for characterizing capsids, but we suggest these conditions might benefit from further optimization with other AAV serotypes or specific PTMs.
Acknowledgements
The authors want to thank Dr. Lass-Napiorkowska of Merck, Process Solutions R&D, Upstream BioProcessing, St. Louis, MO, for providing the AAV5 serotype
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
References
1.
Yan Z, Zak R, Luxton GWG, Ritchie TC, Bantel-Schaal U, Engelhardt JF. 2002. Ubiquitination of both Adeno-Associated Virus Type 2 and 5 Capsid Proteins Affects the Transduction Efficiency of Recombinant Vectors. J Virol. 76(5):2043-2053. https://doi.org/10.1128/jvi.76.5.2043-2053.2002
2.
Mary B, Maurya S, Arumugam S, Kumar V, Jayandharan GR. 2019. Post‐translational modifications in capsid proteins of recombinant adeno‐associated virus (AAV) 1‐rh10 serotypes. The FEBS Journal. 286(24):4964-4981. https://doi.org/10.1111/febs.15013
3.
USP. Analytical Procedures for Viral Vectored Vaccine Quality 2022.
5.
Wu Z, Wang H, Tustian A, Qiu H, Li N. 2022. Development of a Two-Dimensional Liquid Chromatography-Mass Spectrometry Platform for Simultaneous Multi-Attribute Characterization of Adeno-Associated Viruses. Anal. Chem.. 94(7):3219-3226. https://doi.org/10.1021/acs.analchem.1c04873
6.
Wörner TP, Bennett A, Habka S, Snijder J, Friese O, Powers T, Agbandje-McKenna M, Heck AJR. Adeno-associated virus capsid assembly is divergent and stochastic. Nat Commun. 12(1): https://doi.org/10.1038/s41467-021-21935-5
7.
Guapo F, Strasser L, Millán-Martín S, Anderson I, Bones J. 2022. Fast and efficient digestion of adeno associated virus (AAV) capsid proteins for liquid chromatography mass spectrometry (LC-MS) based peptide mapping and post translational modification analysis (PTMs). Journal of Pharmaceutical and Biomedical Analysis. 207114427. https://doi.org/10.1016/j.jpba.2021.114427