Merck
HomeReaction Design & OptimizationAldehydes as Building Blocks
Aldehydes as Building Blocks
Hartmann and co-workers have described the synthesis of a series of potent and selective inhibitors of aldosterone synthase (CYP11B2), in which the key synthetic step was a Wittig reaction using various heterocyclic aldehydes (Scheme 1). The isoquinoline adduct was a potent and selective inhibitor of CYP11B2. The successful inhibition of CYP11B2 has been proposed as a strategy for treatment of congestive heart failure and myocardial fibrosis.1
Aldehydes Scheme
Scheme 1. Product No. 658103 and 675180
4-(1-Pyrrolidino)benzaldehyde was reported as a key building block in the synthesis of an improved inhibitor of NS5B polymerase of the hepatitis C virus (Scheme 2).2 The product from this substrate displayed significant improvement in the potency of the original HTS lead compound (Figure 1).
Aldehydes Scheme
Scheme 2.Product No. 678821
Aldehydes Figure
Figure 1.
Professor Jørgensen’s group reported an asymmetric formal synthesis of (–)-paroxetine, a selective serotonin reuptake inhibitor used in the treatment of anxiety and other psychological disorders.3 The key step involved the organocatalytic conjugate addition of dibenzyl malonate to trans-4-fluorocinnamaldehyde (Scheme 3). Similarly, Wang’s research group has used the Jørgensen organocatalyst in a tandem Michael-aldol reaction to furnish chiral thiochromenes in excellent yields (Scheme 4).4
Aldehydes Scheme
Scheme 3. Product No. 683027
Aldehydes Scheme
Scheme 4. Product No. 683027
Materials
Loading
References
1.
Ulmschneider S, Müller-Vieira U, Klein CD, Antes I, Lengauer T, Hartmann RW. 2005. Synthesis and Evaluation of (Pyridylmethylene)tetrahydronaphthalenes/-indanes and Structurally Modified Derivatives:  Potent and Selective Inhibitors of Aldosterone Synthase. J. Med. Chem.. 48(5):1563-1575. https://doi.org/10.1021/jm0492397
2.
Ding Y, Smith KL, Varaprasad CV, Chang E, Alexander J, Yao N. 2007. Corrigendum to ?Synthesis of thiazolone-based sulfonamides as inhibitors of HCV NS5B polymerase? [Bioorg. Med. Chem. Lett. 17 (2007) 841?845]. Bioorganic & Medicinal Chemistry Letters. 17(9):2665. https://doi.org/10.1016/j.bmcl.2007.03.019
3.
Brandau S, Landa A, Franzén J, Marigo M, Jørgensen KA. 2006. Organocatalytic Conjugate Addition of Malonates to ?,?-Unsaturated Aldehydes: Asymmetric Formal Synthesis of (?)-Paroxetine, Chiral Lactams, and Lactones. Angew. Chem. Int. Ed.. 45(26):4305-4309. https://doi.org/10.1002/anie.200601025
4.
Wang W, Li H, Wang J, Zu L. 2006. Enantioselective Organocatalytic Tandem Michael?Aldol Reactions:  One-Pot Synthesis of Chiral Thiochromenes. J. Am. Chem. Soc.. 128(32):10354-10355. https://doi.org/10.1021/ja063328m