Minimum Essential Medium Eagle (MEM) Formulation
Minimum Essential Medium Eagle (MEM) Alpha Modification
Minimum Essential Medium Eagle (MEM) HEPES Modification
Minimum Essential Medium Eagle (MEM) Joklik Modification for Suspension Cultures
Minimum Essential Medium Eagle (MEM) Modified
Minimum Essential Medium Eagle (MEM) Auto-Mod™ Modified for Autoclaving
Minimum Essential Medium Eagle (S-MEM) Spinner Modification
Minimum Essential Medium Eagle (MEM) | |||||
---|---|---|---|---|---|
Component | M0268 g/L | M0275 [10x] g/L | M0446 [1x] g/L | M0643 g/L | M1018 g/L |
Inorganic Salts | |||||
Calcium Chloride | 0.2 | 2 | 0.2 | 0.2 | 0.1396 |
Magnesium Sulfate (anhydrous) | 0.09767 | 0.9767 | 0.09767 | 0.09767 | 0.09767 |
Potassium Chloride | 0.4 | 4 | 0.4 | 0.4 | 0.4 |
Potassium Phosphate Monobasic (anhydrous) | — | — | — | — | 0.06 |
Sodium Bicarbonate | — | — | 2.2 | — | — |
Sodium Chloride | 6.8 | 68 | 6.8 | 6.8 | 8 |
Sodium Phosphate Dibasic (anhydrous) | — | — | — | — | 0.04788 |
Sodium Phosphate Monobasic (anhydrous) | 0.122 | 1.22 | 0.122 | 0.122 | — |
Amino Acids | |||||
L-Alanine | — | — | — | 0.0089 | 0.0089 |
L-Alanyl-L-Glutamine | — | — | 0.4344 | — | — |
L-Arginine • HCl | 0.126 | 1.26 | 0.126 | 0.126 | 0.126 |
L-Asparagine • H2O | — | — | — | 0.015 | 0.015 |
L-Aspartic Acid | — | — | — | 0.0133 | 0.0133 |
L-Cystine • 2HCl | 0.0313 | 0.313 | 0.0313 | 0.0313 | 0.0313 |
L-Glutamic Acid | — | — | — | 0.0147 | 0.0147 |
L-Glutamine | 0.292 | — | 0.292 | 0.292 | 0.292 |
Glycine | — | — | — | 0.0075 | 0.0075 |
L-Histidine • HCl • H2O | 0.042 | 0.42 | 0.042 | 0.042 | 0.042 |
L-Isoleucine | 0.052 | 0.52 | 0.052 | 0.052 | 0.052 |
L-Leucine | 0.052 | 0.52 | 0.052 | 0.052 | 0.052 |
L-Lysine • HCl | 0.0725 | 0.725 | 0.0725 | 0.0725 | 0.0725 |
L-Methionine | 0.015 | 0.15 | 0.015 | 0.015 | 0.015 |
L-Phenylalanine | 0.032 | 0.32 | 0.032 | 0.032 | 0.032 |
L-Proline | — | — | — | 0.0115 | 0.0115 |
L-Serine | — | — | — | 0.0105 | 0.0105 |
L-Threonine | 0.048 | 0.48 | 0.048 | 0.048 | 0.048 |
L-Tryptophan | 0.01 | 0.1 | 0.01 | 0.01 | 0.01 |
L-Tyrosine • 2Na • 2H2O | 0.0519 | 0.519 | 0.0519 | 0.0519 | 0.0519 |
L-Valine | 0.046 | 0.46 | 0.046 | 0.046 | 0.046 |
Vitamins | |||||
Choline Chloride | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 |
Folic Acid | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 |
myo-Inositol | 0.002 | 0.02 | 0.002 | 0.002 | 0.002 |
Niacinamide | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 |
Pyridoxal • HCl | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 |
Riboflavin | 0.0001 | 0.001 | 0.0001 | 0.0001 | 0.0001 |
Thiamine • HCl | 0.001 | 0.01 | 0.001 | 0.001 | 0.001 |
Other | |||||
Glucose | 1 | 10 | 1 | 1 | 1 |
Phenol Red • Na | 0.011 | 0.11 | 0.011 | 0.011 | 0.011 |
Add | |||||
L-Glutamine | — | 0.292 at 1× | — | — | — |
Sodium Bicarbonate | 2.2 | 2.2 at 1× | — | 2.2 | 0.35 |
Minimum Essential Medium Eagle (MEM) (cont.) | ||||
---|---|---|---|---|
Component | M2279 g/L | M2414 [1x] g/L | M4642 [1x] g/L | M4655 g/L |
Inorganic Salts | ||||
Calcium Chloride | 0.2 | 0.2 | 0.2 | 0.2 |
Magnesium Sulfate (anhydrous) | 0.09767 | 0.09767 | 0.09767 | 0.09767 |
Potassium Chloride | 0.4 | 0.4 | 0.4 | 0.4 |
Potassium Phosphate Monobasic (anhydrous) | — | — | 0.06 | — |
Sodium Bicarbonate | 2.2 | 2.2 | — | 2.2 |
Sodium Chloride | 6.8 | 6.8 | 8 | 6.8 |
Sodium Phosphate Dibasic (anhydrous) | — | — | 0.04788 | — |
Sodium Phosphate Monobasic (anhydrous) | 0.122 | 0.122 | — | 0.122 |
Amino Acids | ||||
L-Alanine | — | — | — | — |
L-Arginine • HCl | 0.126 | 0.126 | 0.126 | 0.126 |
L-Asparagine • H2O | — | — | — | — |
L-Aspartic Acid | — | — | — | — |
L-Cystine • 2HCl | 0.0313 | 0.0313 | 0.0313 | 0.0313 |
L-Glutamic Acid | — | — | — | — |
L-Glutamine | — | 0.292 | 0.292 | 0.292 |
Glycine | — | — | — | — |
L-Histidine • HCl • H2O | 0.042 | 0.042 | 0.042 | 0.042 |
L-Isoleucine | 0.052 | 0.052 | 0.052 | 0.052 |
L-Leucine | 0.052 | 0.052 | 0.052 | 0.052 |
L-Lysine • HCl | 0.0725 | 0.0725 | 0.0725 | 0.0725 |
L-Methionine | 0.015 | 0.015 | 0.015 | 0.015 |
L-Phenylalanine | 0.032 | 0.032 | 0.032 | 0.032 |
L-Proline | — | — | — | — |
L-Serine | — | — | — | — |
L-Threonine | 0.048 | 0.048 | 0.048 | 0.048 |
L-Tryptophan | 0.01 | 0.01 | 0.01 | 0.01 |
L-Tyrosine • 2Na • 2H2O | 0.0519 | 0.0519 | 0.0519 | 0.0519 |
L-Valine | 0.046 | 0.046 | 0.046 | 0.046 |
Vitamins | ||||
Choline Chloride | 0.001 | 0.001 | 0.001 | 0.001 |
Folic Acid | 0.001 | 0.001 | 0.001 | 0.001 |
myo-Inositol | 0.002 | 0.002 | 0.002 | 0.002 |
Niacinamide | 0.001 | 0.001 | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 | 0.001 | 0.001 | 0.001 |
Pyridoxal • HCl | 0.001 | 0.001 | 0.001 | 0.001 |
Riboflavin | 0.0001 | 0.0001 | 0.0001 | 0.0001 |
Thiamine • HCl | 0.001 | 0.001 | 0.001 | 0.001 |
Other | ||||
Glucose | 1 | 1 | 1 | 1 |
Phenol Red • Na | 0.011 | 0.011 | 0.011 | 0.011 |
Add | ||||
L-Glutamine | 0.292 | — | — | — |
Sodium Bicarbonate | — | — | 0.35 | — |
Minimum Essential Medium Eagle (MEM) (cont.) | ||||
---|---|---|---|---|
Component | M4780 [1x] g/L | M5650 [1x] g/L | M5775 [1x] g/L | M9288 [10x] g/L |
Inorganic Salts | ||||
Calcium Chloride | 0.1396 | 0.2 | 0.1396 | 1.396 |
Magnesium Sulfate (anhydrous) | 0.09767 | 0.09767 | 0.09767 | 0.9767 |
Potassium Chloride | 0.4 | 0.4 | 0.4 | 4 |
Potassium Phosphate Monobasic (anhydrous) | 0.06 | — | 0.06 | 0.6 |
Sodium Bicarbonate | 0.35 | 2.2 | 0.35 | — |
Sodium Chloride | 8 | 6.8 | 8 | 80 |
Sodium Phosphate Dibasic (anhydrous) | 0.04788 | — | 0.04788 | 0.4788 |
Sodium Phosphate Monobasic (anhydrous) | — | 0.122 | — | — |
Amino Acids | ||||
L-Alanine | — | 0.0089 | — | — |
L-Arginine • HCl | 0.126 | 0.126 | 0.126 | 1.26 |
L-Asparagine • H2O | — | 0.015 | — | — |
L-Aspartic Acid | — | 0.0133 | — | — |
L-Cystine • 2HCl | 0.0313 | 0.0313 | 0.0313 | 0.313 |
L-Glutamic Acid | — | 0.0147 | — | — |
L-Glutamine | 0.292 | — | — | — |
Glycine | — | 0.0075 | — | — |
L-Histidine • HCl • H2O | 0.042 | 0.042 | 0.042 | 0.42 |
L-Isoleucine | 0.052 | 0.052 | 0.052 | 0.52 |
L-Leucine | 0.052 | 0.052 | 0.052 | 0.52 |
L-Lysine • HCl | 0.0725 | 0.0725 | 0.0725 | 0.725 |
L-Methionine | 0.015 | 0.015 | 0.015 | 0.15 |
L-Phenylalanine | 0.032 | 0.032 | 0.032 | 0.32 |
L-Proline | — | 0.0115 | — | — |
L-Serine | — | 0.0105 | — | — |
L-Threonine | 0.048 | 0.048 | 0.048 | 0.48 |
L-Tryptophan | 0.01 | 0.01 | 0.01 | 0.1 |
L-Tyrosine • 2Na • 2H2O | 0.0519 | 0.0519 | 0.0519 | 0.519 |
L-Valine | 0.046 | 0.046 | 0.046 | 0.46 |
Vitamins | ||||
Choline Chloride | 0.001 | 0.001 | 0.001 | 0.01 |
Folic Acid | 0.001 | 0.001 | 0.001 | 0.01 |
myo-Inositol | 0.002 | 0.002 | 0.002 | 0.02 |
Niacinamide | 0.001 | 0.001 | 0.001 | 0.01 |
D-Pantothenic Acid (hemicalcium) | 0.001 | 0.001 | 0.001 | 0.01 |
Pyridoxal • HCl | 0.001 | 0.001 | 0.001 | 0.01 |
Riboflavin | 0.0001 | 0.0001 | 0.0001 | 0.001 |
Thiamine • HCl | 0.001 | 0.001 | 0.001 | 0.01 |
Other | ||||
Glucose | 1 | 1 | 1 | 10 |
Phenol Red • Na | 0.011 | 0.011 | 0.011 | 0.11 |
Add | ||||
L-Glutamine | — | 0.292 | 0.292 | 0.292 at 1× |
Sodium Bicarbonate | — | — | — | 0.35 at 1× |
Minimum Essential Medium Eagle (MEM) Alpha Modification | ||||
---|---|---|---|---|
Component | M0644 g/L | M0894 g/L | M4526 [1x] g/L | M8042 [1x] g/L |
Inorganic Salts | ||||
Calcium Chloride | 0.2 | 0.2 | 0.2 | 0.2 |
Magnesium Sulfate (anhydrous) | 0.09767 | 0.09767 | 0.09767 | 0.09767 |
Potassium Chloride | 0.4 | 0.4 | 0.4 | 0.4 |
Sodium Bicarbonate | — | — | 2.2 | 2.2 |
Sodium Chloride | 6.8 | 6.8 | 6.8 | 6.8 |
Sodium Phosphate Monobasic (anhydrous) | 0.122 | 0.122 | 0.122 | 0.122 |
Amino Acids | ||||
L-Alanine | 0.025 | 0.025 | 0.025 | 0.025 |
L-Arginine • HCl | 0.126 | 0.126 | 0.126 | 0.126 |
L-Asparagine • H2O | 0.05 | 0.05 | 0.05 | 0.05 |
L-Aspartic Acid | 0.03 | 0.03 | 0.03 | 0.03 |
L-Cysteine • HCl • H2O | 0.1 | 0.1 | 0.1 | 0.1 |
L-Cystine • 2HCl | 0.0313 | 0.0313 | 0.0313 | 0.0313 |
L-Glutamic Acid | 0.075 | 0.075 | 0.075 | 0.075 |
L-Glutamine | 0.292 | 0.292 | — | — |
Glycine | 0.05 | 0.05 | 0.05 | 0.05 |
L-Histidine • HCl • H2O | 0.042 | 0.042 | 0.042 | 0.042 |
L-Isoleucine | 0.052 | 0.052 | 0.052 | 0.052 |
L-Leucine | 0.052 | 0.052 | 0.052 | 0.052 |
L-Lysine • HCl | 0.0725 | 0.0725 | 0.0725 | 0.0725 |
L-Methionine | 0.015 | 0.015 | 0.015 | 0.015 |
L-Phenylalanine | 0.032 | 0.032 | 0.032 | 0.032 |
L-Proline | 0.04 | 0.04 | 0.04 | 0.04 |
L-Serine | 0.025 | 0.025 | 0.025 | 0.025 |
L-Threonine | 0.048 | 0.048 | 0.048 | 0.048 |
L-Tryptophan | 0.01 | 0.01 | 0.01 | 0.01 |
L-Tyrosine • 2Na • 2H2O | 0.0519 | 0.0519 | 0.0519 | 0.0519 |
L-Valine | 0.046 | 0.046 | 0.046 | 0.046 |
Vitamins | ||||
L-Ascorbic Acid • Na | 0.05 | 0.05 | 0.05 | 0.05 |
Biotin | 0.0001 | 0.0001 | 0.0001 | 0.0001 |
Choline Chloride | 0.001 | 0.001 | 0.001 | 0.001 |
Folic Acid | 0.001 | 0.001 | 0.001 | 0.001 |
myo-Inositol | 0.002 | 0.002 | 0.002 | 0.002 |
Lipoic Acid | — | — | 0.0002 | — |
Niacinamide | 0.001 | 0.001 | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 | 0.001 | 0.001 | 0.001 |
Pyridoxal • HCl | 0.001 | 0.001 | 0.001 | 0.001 |
Riboflavin | 0.0001 | 0.0001 | 0.0001 | 0.0001 |
Thiamine • HCl | 0.001 | 0.001 | 0.001 | 0.001 |
Vitamin B12 | 0.00136 | 0.00136 | 0.00136 | 0.00136 |
Other | ||||
Adenosine | 0.01 | — | — | 0.01 |
Cytidine | 0.01 | — | — | 0.01 |
2'-Deoxyadenosine | 0.01 | — | — | 0.01 |
2'-Deoxycytidine • HCl | 0.011 | — | — | 0.011 |
2'-Deoxyguanosine | 0.01 | — | — | 0.01 |
Glucose | 1 | 1 | 1 | 1 |
Guanosine | 0.01 | — | — | 0.01 |
Phenol Red • Na | 0.011 | 0.011 | 0.011 | 0.011 |
Pyruvic Acid • Na | 0.11 | 0.11 | 0.11 | 0.11 |
Thioctic Acid | 0.0002 | 0.0002 | — | 0.0002 |
Thymidine | 0.01 | — | — | 0.01 |
Uridine | 0.01 | — | — | 0.01 |
Add | ||||
L-Glutamine | — | — | 0.292 | 0.292 |
Sodium Bicarbonate | 2.2 | 2.2 | — | — |
Minimum Essential Medium Eagle (MEM) HEPES Modification | ||
---|---|---|
Component | M2645 g/L | M7278 [1x] g/L |
Inorganic Salts | ||
Calcium Chloride | 0.2 | 0.2 |
Magnesium Sulfate (anhydrous) | 0.09767 | 0.09767 |
Potassium Chloride | 0.4 | 0.4 |
Sodium Bicarbonate | — | 2.2 |
Sodium Chloride | 5.5 | 5.5 |
Sodium Phosphate Monobasic (anhydrous) | 0.122 | 0.122 |
Amino Acids | ||
L-Arginine • HCl | 0.126 | 0.126 |
L-Cystine • 2HCl | 0.0313 | 0.0313 |
L-Glutamine | 0.292 | — |
L-Histidine • HCl • H2O | 0.042 | 0.042 |
L-Isoleucine | 0.052 | 0.052 |
L-Leucine | 0.052 | 0.052 |
L-Lysine • HCl | 0.0725 | 0.0725 |
L-Methionine | 0.015 | 0.015 |
L-Phenylalanine | 0.032 | 0.032 |
L-Threonine | 0.048 | 0.048 |
L-Tryptophan | 0.01 | 0.01 |
L-Tyrosine • 2Na • 2H2O | 0.0519 | 0.0519 |
L-Valine | 0.046 | 0.046 |
Vitamins | ||
Choline Chloride | 0.001 | 0.001 |
Folic Acid | 0.001 | 0.001 |
myo-Inositol | 0.002 | 0.002 |
Niacinamide | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 | 0.001 |
Pyridoxal • HCl | 0.001 | 0.001 |
Riboflavin | 0.0001 | 0.0001 |
Thiamine • HCl | 0.001 | 0.001 |
Other | ||
Glucose | 1 | 1 |
HEPES | 5.958 | 5.958 |
Phenol Red • Na | 0.011 | 0.011 |
Add | ||
L-Glutamine | — | 0.292 |
Sodium Bicarbonate | 2.2 | — |
Minimum Essential Medium Eagle (MEM) Joklik Modification for Suspension Cultures | ||
---|---|---|
Component | M0518 g/L | M8028 [1x] g/L |
Inorganic Salts | ||
Magnesium Chloride • 6H2O | 0.2 | 0.2 |
Potassium Chloride | 0.4 | 0.4 |
Sodium Bicarbonate | — | 2 |
Sodium Chloride | 6.5 | 6.5 |
Sodium Phosphate Monobasic (anhydrous) | 1.154 | 1.154 |
Amino Acids | ||
L-Arginine • HCl | 0.126 | 1.26 |
L-Cystine • 2HCl | 0.0324 | 0.0324 |
L-Glutamine | 0.292 | — |
L-Histidine • HCl • H2O | 0.042 | 0.042 |
L-Isoleucine | 0.052 | 0.052 |
L-Leucine | 0.052 | 0.052 |
L-Lysine • HCl | 0.0725 | 0.0725 |
L-Methionine | 0.015 | 0.015 |
L-Phenylalanine | 0.032 | 0.032 |
L-Threonine | 0.048 | 0.048 |
L-Tryptophan | 0.01 | 0.01 |
L-Tyrosine • 2Na • 2H2O | 0.05452 | 0.05452 |
L-Valine | 0.046 | 0.046 |
Vitamins | ||
Choline Chloride | 0.001 | 0.001 |
Folic Acid | 0.001 | 0.001 |
myo-Inositol | 0.002 | 0.002 |
Niacinamide | 0.001 | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 | 0.001 |
Pyridoxal • HCl | 0.001 | 0.001 |
Riboflavin | 0.0001 | 0.0001 |
Thiamine • HCl | 0.001 | 0.001 |
Other | ||
Glucose | 2 | 2 |
Phenol Red • Na | 0.011 | 0.011 |
Add | ||
L-Glutamine | — | 0.292 |
Sodium Bicarbonate | 2 | — |
Minimum Essential Medium Eagle (MEM) Modified | |
---|---|
Component | M3024 g/L |
Inorganic Salts | |
Calcium Chloride | 0.2 |
Magnesium Sulfate (anhydrous) | 0.09767 |
Potassium Chloride | 0.4 |
Sodium Chloride | 6.8 |
Sodium Phosphate Monobasic (anhydrous) | 0.122 |
Amino Acids | |
L-Alanine | 0.0089 |
L-Arginine • HCl | 0.126 |
L-Asparagine • H2O | 0.015 |
L-Aspartic Acid | 0.0133 |
L-Cystine • 2HCl | 0.0313 |
L-Glutamic Acid | 0.0147 |
Glycine | 0.0075 |
L-Histidine • HCl • H2O | 0.042 |
L-Isoleucine | 0.052 |
L-Leucine | 0.052 |
L-Lysine • HCl | 0.0725 |
L-Methionine | 0.015 |
L-Phenylalanine | 0.032 |
L-Proline | 0.0115 |
L-Serine | 0.0105 |
L-Threonine | 0.048 |
L-Tryptophan | 0.01 |
L-Tyrosine • 2Na • 2H2O | 0.0519 |
L-Valine | 0.046 |
Vitamins | |
Choline Chloride | 0.001 |
Folic Acid | 0.001 |
myo-Inositol | 0.002 |
Niacinamide | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 |
Pyridoxal • HCl | 0.001 |
Riboflavin | 0.0001 |
Thiamine • HCl | 0.001 |
Other | |
Glucose | 1 |
Add | |
L-Glutamine | 0.292 |
Sodium Bicarbonate | 2.2 |
Minimum Essential Medium Eagle (MEM) Auto-Mod™ Modified for Autoclaving | |
---|---|
Component | M0769 g/L |
Inorganic Salts | |
Calcium Chloride | 0.2 |
Magnesium Sulfate (anhydrous) | 0.09767 |
Potassium Chloride | 0.4 |
Sodium Chloride | 6.8 |
Sodium Phosphate Monobasic (anhydrous) | 0.122 |
Sodium Succinate • 6H2O | 0.1 |
Succinic Acid (free acid) | 0.075 |
Amino Acids | |
L-Arginine • HCl | 0.126 |
L-Cystine • 2HCl | 0.0313 |
L-Histidine • HCl • H2O | 0.042 |
L-Isoleucine | 0.052 |
L-Leucine | 0.052 |
L-Lysine • HCl | 0.0725 |
L-Methionine | 0.015 |
L-Phenylalanine | 0.032 |
L-Threonine | 0.048 |
L-Tryptophan | 0.01 |
L-Tyrosine (free base) | 0.036 |
L-Valine | 0.046 |
Vitamins | |
Choline Bitartrate | 0.0018 |
Folic Acid | 0.001 |
myo-Inositol | 0.002 |
Niacinamide | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 |
Pyridoxal • HCl | 0.001 |
Riboflavin | 0.0001 |
Thiamine • HCl | 0.001 |
Other | |
Glucose | 1 |
Phenol Red • Na | 0.0064 |
Add | |
L-Glutamine | 0.292 |
Sodium Bicarbonate | 2.2 |
Minimum Essential Medium Eagle (S-MEM) Spinner Modification | |
---|---|
Component | M8167 [1x] g/L |
Inorganic Salts | |
Magnesium Sulfate (anhydrous) | 0.09767 |
Potassium Chloride | 0.4 |
Sodium Chloride | 6.8 |
Sodium Phosphate Monobasic (anhydrous) | 0.122 |
Amino Acids | |
L-Arginine • HCl | 0.126 |
L-Cystine • 2HCl | 0.0313 |
L-Histidine • HCl • H2O | 0.042 |
L-Isoleucine | 0.052 |
L-Leucine | 0.052 |
L-Lysine • HCl | 0.0725 |
L-Methionine | 0.015 |
L-Phenylalanine | 0.032 |
L-Threonine | 0.048 |
L-Tryptophan | 0.01 |
L-Tyrosine • 2Na • 2H2O | 0.0519 |
L-Valine | 0.046 |
Vitamins | |
Choline Chloride | 0.001 |
Folic Acid | 0.001 |
myo-Inositol | 0.002 |
Niacinamide | 0.001 |
D-Pantothenic Acid (hemicalcium) | 0.001 |
Pyridoxal • HCl | 0.001 |
Riboflavin | 0.0001 |
Thiamine • HCl | 0.001 |
Other | |
Glucose | 1 |
Phenol Red • Na | 0.011 |
Sodium Bicarbonate | 2.2 |
Add | |
L-Glutamine | 0.292 |