Merck
UHPLC Analysis of Diazepam and Metabolites on Titan C18, Competitive Comparison
UHPLC Analysis of Diazepam and Metabolites on Titan C18, Competitive Comparison application for UHPLC
Materials
standard
Product No.
Description
Pricing
Diazepam
Uracil
≥99.0%
Oxazepam
Desmethyldiazepam
analytical standard
Temazepam
analytical standard
Methyl benzoate
analytical standard
CONDITIONS
column
Titan C18, 10 cm x 2.1 mm I.D., 1.9 μm particles (577124-U)
mobile phase
[A] 0.1% formic acid; [B] acetonitrile; (60:40, A:B)
pressure
6450 psi (445 bar)
column temp.
35 °C
detector
UV, 254 nm
Description
Analysis Note
This application shows a comparison between Titan C18 and a competitive column for diazepam and its metabolites using a MS compatible mobile phase.
Other Notes
Used 75 μm I.D. x 250 mm L tubing in system.
Legal Information
null