Merck
Higher Efficiency of Ascentis® Express 2.7 μm Compared to 3 μm Particles Gives Better Sensitivity
Higher Efficiency of Ascentis® Express 2.7 μm Compared to 3 μm Particles Gives Better Sensitivity application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 5 cm × 2.1 mm
CONDITIONS
column
Ascentis Express C18, 5 cm x 2.1 mm I.D., 2.7 μm particles and C18, 5 cm x 2.1 mm I.D., 3 μm particles (53822-U)
mobile phase
[A] 25 mM dibasic ammonium phosphate (pH 7.0): [B] water: [C] acetonitrile; (35:0:65 or 35:4:61, A:B:C)
flow rate
0.2 mL/min
column temp.
35 °C
detector
UV, 220 nm
injection
1 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany