HPLC Analysis of Nicardipine Enantiomers on Astec® CHIROBIOTIC® V
![HPLC Analysis of Nicardipine Enantiomers on Astec® CHIROBIOTIC® V application for HPLC](/deepweb/assets/sigmaaldrich/product/images/768/308/e93ac8c0-b401-4a24-af31-da1663ae8a63/800/e93ac8c0-b401-4a24-af31-da1663ae8a63.jpg)
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine; (100:0.02:0.01, A:B:C)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 254 nm
injection
2 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC