GC Analysis of Spearmint Essential Oil on SLB®-IL59, 80 °C Initial Oven Temp.
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL59, 30 m x 0.25 mm I.D., 0.20 μm (28891-U)
oven
80 °C, 3 °C/min to 280 °
inj. temp.
250 °C
detector
FID, 305 °C
carrier gas
helium, 40 cm/sec
injection
1 μL 100:1 split
sample
spearmint essential oil diluted 1:10 in hexane
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany