Merck
GC Analysis of Bupivacaine Enantiomers on Astec® CHIRALDEX B-PH
GC Analysis of Bupivacaine Enantiomers on Astec® CHIRALDEX™ B-PH suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX B-PH Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
Bupivacaine hydrochloride monohydrate
analytical standard, for drug analysis
CONDITIONS
column
Astec CHIRALDEX B-PH, 10 m x 0.25 mm I.D., 0.12 μm (custom)
gradient
130 °C, 2 °C/min to 170 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 12 psi
sample
peaks 1 & 2: bupivacaine enantiomers
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC