US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Materials
analytical column
accessory
Wheaton wide-mouth bottle
amber soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 eaguard column
standard
EPA 8270 GC-MS Tuning Solution II
certified reference material, 1000 μg/mL each component in dichloromethaneEPA CLP Semivolatile Calibration Mix
certified reference material, 1000 μg/mL each component in dichloromethane : benzene (3:1)EPA CLP Semivolatiles Surrogate Standards Mix
certified reference material, 4000 μg/mL each component in methylene chlorideEPA 8270 Semivolatile Internal Standard Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methylene chloride, ampule of 2 × 1 mLEPA 8270 Benzidines Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methanol, ampule of 1 mLCONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.50 μm (28473-U)
oven
40 °C (1 min), 12 °C/min to 250 °C, 25 °C/min to 340 °C (4.5 min)
inj. temp.
250 °C
MSD interface
340 °C
scan range
m/z = 40-450
carrier gas
helium, 1.5 mL/min constant flow
injection
1.0 μL, splitless (0.50 min)
liner
4 mm I.D., single taper
sample
50 ng on-column of a 72 component semivolatile standard and 8 surrogate compounds, plus 6 internal standards (at 40 ng on-column)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany