US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap
Materials
analytical column
CONDITIONS
sample
10 ppb each component in 5 mL water
purge trap
VOCARB 3000 (21066-U)
purge
40 mL/min for 11 min
dry purge
3 min
desorption process
250 °C for 4 min
bake
280 °C for 10 min
column
VOCOL, 105 m × 0.53 mm I.D., 3.0 μm (25358)
oven
35 °C (10 min), 4 °C/min to 200 °C (10 min)
scan range
m/z = 35-260 at 0.6 scan/sec
carrier gas
helium, 10 mL/min
Description
Legal Information
VOCOL is a registered trademark of Merck KGaA, Darmstadt, Germany