Merck
  • Hinokitiol inhibits compound action potentials in the frog sciatic nerve.
Hinokitiol inhibits compound action potentials in the frog sciatic nerve.
European journal of pharmacology (2017-12-12)
Nobuya Magori, Tsugumi Fujita, Eiichi Kumamoto
ABSTRACT
Hinokitiol (β-thujaplicin) is a natural tropolone derivative contained in Chamaecyparis taiwanensis that has various actions including anti-inflammatory activities. Various plant-derived compounds inhibit compound action potentials (CAPs) in a manner dependent on the chemical structure of the compounds; however, the effects of hinokitiol on nerve conduction have not been examined. To determine whether hinokitiol inhibits CAPs and, if so, the chemical structure of hinokitiol that is important in this inhibition, we examined the effects of hinokitiol and its related compounds on fast-conducting CAPs using the frog sciatic nerve and the air-gap method. Hinokitiol concentration-dependently reduced the peak amplitude of CAPs with a half-maximal inhibitory concentration (IC
Sigma-Aldrich
Guaiazulene, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Isopropyl-3-methylphenol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Isopropylphenol, 98%
Pricing and availability is not currently available.