Merck
A Novel Role for
Frontiers in genetics (2018-01-13)
Amina Kamar, Akl C Fahed, Kamel Shibbani, Nehme El-Hachem, Salim Bou-Slaiman, Mariam Arabi, Mazen Kurban, Jonathan G Seidman, Christine E Seidman, Rachid Haidar, Elias Baydoun, Georges Nemer, Fadi Bitar
ABSTRACT
Despite an obvious role for consanguinity in congenital heart disease (CHD), most studies fail to document a monogenic model of inheritance except for few cases. We hereby describe a first-degree cousins consanguineous Lebanese family with 7 conceived children: 2 died
Sigma-Aldrich
StableCell DMEM - high glucose, With 4500 mg/L glucose, stable glutamine, and sodium bicarbonate, without sodium pyruvate., liquid, sterile-filtered, suitable for cell culture