Merck
Does
Veterinary research forum : an international quarterly journal (2017-08-09)
Mojtaba Mohseni, Masoud Maham, Bahram Dalir-Naghadeh, Ghader Jalilzadeh-Amin
ABSTRACT
Displacement of the abomasum is a common disease of the gastrointestinal tract in dairy cattle. Abomasal displacement has been associated with abomasal hypomotility. Therefore, it is necessary to identify effective therapeutic agents that stimulate abomasal motility in cattle. Yarrow (
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
4-DAMP, ≥98% (HPLC)