Merck
  • Magnesium-selective ion-channel mimetic sensor with a traditional calcium ionophore.
Magnesium-selective ion-channel mimetic sensor with a traditional calcium ionophore.
Analytical chemistry (2009-12-04)
Jingwei Zhu, Yu Qin, Yunhong Zhang
ABSTRACT
A novel magnesium-selective ion-channel mimetic sensor based on a well-known calcium ionophore, ETH 5234, was demonstrated. Due to different stoichiometry of the ionophore with Mg(2+) and Ca(2+), the sensor exhibits high selectivity to Mg(2+) by controlling ionophore concentration on the electrode surface, which allows the measurement of Mg(2+) in a 1 mM Ca(2+) background solution.
Product Description
Supelco
Calcium ionophore IV, for ion-selective electrodes, Selectophore, function tested
Pricing and availability is not currently available.