Merck
  • SHARPIN-mediated regulation of protein arginine methyltransferase 5 controls melanoma growth.
SHARPIN-mediated regulation of protein arginine methyltransferase 5 controls melanoma growth.
The Journal of clinical investigation (2017-12-12)
Hironari Tamiya, Hyungsoo Kim, Oleksiy Klymenko, Heejung Kim, Yongmei Feng, Tongwu Zhang, Ji Yun Han, Ayako Murao, Scott J Snipas, Lucia Jilaveanu, Kevin Brown, Harriet Kluger, Hao Zhang, Kazuhiro Iwai, Ze'ev A Ronai
ABSTRACT
SHARPIN, an adaptor for the linear ubiquitin chain assembly complex (LUBAC), plays important roles in NF-κB signaling and inflammation. Here, we have demonstrated a LUBAC-independent role for SHARPIN in regulating melanoma growth. We observed that SHARPIN interacted with PRMT5, a type II protein arginine methyltransferase, and increased its multiprotein complex and methyltransferase activity. Activated PRMT5 controlled the expression of the transcription factors SOX10 and MITF by SHARPIN-dependent arginine dimethylation and inhibition of the transcriptional corepressor SKI. Activation of PRMT5 by SHARPIN counteracted PRMT5 inhibition by methylthioadenosine, a substrate of methylthioadenosine phosphorylase, which is codeleted with cyclin-dependent kinase inhibitor 2A (CDKN2A) in approximately 15% of human cancers. Collectively, we identified a LUBAC-independent role for SHARPIN in enhancing PRMT5 activity that contributes to melanomagenesis through the SKI/SOX10 regulatory axis.
Sigma-Aldrich
Anti-dimethyl-Arginine Antibody, symmetric (SYM10), serum, Upstate®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)