Merck
  • Hydrothermal treatment and butylphosphonic acid derived self-assembled monolayers for improving the surface chemistry and corrosion resistance of AZ61 magnesium alloy.
Hydrothermal treatment and butylphosphonic acid derived self-assembled monolayers for improving the surface chemistry and corrosion resistance of AZ61 magnesium alloy.
Scientific reports (2017-12-06)
Chung-Wei Yang, Cheng Liu, Da-Jun Lin, Ming-Long Yeh, Tzer-Min Lee
ABSTRACT
The hydrothermal treatment followed by a self-assembled monolayer (SAM) of 1-butylphosphonic acid through the tethering by aggregation and growth (T-BAG) method was employed to produce protective surface coatings on the Mg-6Al-1Zn alloy (AZ61) for reducing the degradation rate in physiological environments. Potentiodynamic polarization measurements revealed that the organic self-assembled monolayer and Mg(OH)
Sigma-Aldrich
Butylphosphonic acid