Merck
  • Staphylococcus aureus produces pain through pore-forming toxins and neuronal TRPV1 that is silenced by QX-314.
Staphylococcus aureus produces pain through pore-forming toxins and neuronal TRPV1 that is silenced by QX-314.
Nature communications (2018-01-04)
Kimbria J Blake, Pankaj Baral, Tiphaine Voisin, Ashira Lubkin, Felipe Almeida Pinho-Ribeiro, Kelsey L Adams, David P Roberson, Yuxin C Ma, Michael Otto, Clifford J Woolf, Victor J Torres, Isaac M Chiu
ABSTRACT
The hallmark of many bacterial infections is pain. The underlying mechanisms of pain during live pathogen invasion are not well understood. Here, we elucidate key molecular mechanisms of pain produced during live methicillin-resistant Staphylococcus aureus (MRSA) infection. We show that spontaneous pain is dependent on the virulence determinant agr and bacterial pore-forming toxins (PFTs). The cation channel, TRPV1, mediated heat hyperalgesia as a distinct pain modality. Three classes of PFTs-alpha-hemolysin (Hla), phenol-soluble modulins (PSMs), and the leukocidin HlgAB-directly induced neuronal firing and produced spontaneous pain. From these mechanisms, we hypothesized that pores formed in neurons would allow entry of the membrane-impermeable sodium channel blocker QX-314 into nociceptors to silence pain during infection. QX-314 induced immediate and long-lasting blockade of pain caused by MRSA infection, significantly more than lidocaine or ibuprofen, two widely used clinical analgesic treatments.
Sigma-Aldrich
Lidocaine N-ethyl bromide, analytical standard, for drug analysis
Pricing and availability is not currently available.