MilliporeSigma
Sample processing approach for detection of ricin in surface samples.
Journal of immunological methods (2017-09-01)
Staci Kane, Sanjiv Shah, Anne Marie Erler, Teneile Alfaro
ABSTRACT
With several ricin contamination incidents reported over the past decade, rapid and accurate methods are needed for environmental sample analysis, especially after decontamination. A sample processing method was developed for common surface sampling devices to improve the limit of detection and avoid false negative/positive results for ricin analysis. Potential assay interferents from the sample matrix (bleach residue, sample material, wetting buffer), including reference dust, were tested using a Time-Resolved Fluorescence (TRF) immunoassay. Test results suggested that the sample matrix did not cause the elevated background fluorescence sometimes observed when analyzing post-bleach decontamination samples from ricin incidents. Furthermore, sample particulates (80mg/mL Arizona Test Dust) did not enhance background fluorescence or interfere with ricin detection by TRF. These results suggested that high background fluorescence in this immunoassay could be due to labeled antibody quality and/or quantity issues. Centrifugal ultrafiltration devices were evaluated for ricin concentration as a part of sample processing. Up to 30-fold concentration of ricin was observed by the devices, which serve to remove soluble interferents and could function as the front-end sample processing step to other ricin analytical methods. The procedure has the potential to be used with a broader range of environmental sample types and with other potential interferences and to be followed by other ricin analytical methods, although additional verification studies would be required.