Merck
  • Beyond Born-Rytov limit for super-resolution optical diffraction tomography.
Beyond Born-Rytov limit for super-resolution optical diffraction tomography.
Optics express (2017-12-10)
JooWon Lim, Abdul Wahab, GwangSik Park, KyeoReh Lee, YongKeun Park, Jong Chul Ye
ABSTRACT
Optical diffraction tomography (ODT) using Born or Rytov approximation suffers from severe distortions in reconstructed refractive index (RI) tomograms when multiple scattering occurs or the scattering signals are strong. These effects are usually seen as a significant impediment to the application of ODT because multiple scattering is directly linked to an unknown object itself rather than a surrounding medium, and a strong scatter invalidates the underlying assumptions of the Born and Rytov approximations. The focus of this article is to demonstrate for the first time that multiple scattering and high material contrast, if handled aptly, can significantly improve the image quality of the ODT thanks to multiple scattering inside a sample. Experimental verification using various phantom and biological cells substantiates that we not only revealed the structures that were not observable using the conventional approaches but also resolved the long-standing problem of missing cones in the ODT.
Product Description
Supelco
Micro particle size standard based on polystyrene monodisperse, size: 0.2 μm
Pricing and availability is not currently available.
Supelco
Micro particles based on silicon dioxide, size: 5 μm
Pricing and availability is not currently available.