Merck
  • Multimodal Theranostic Nanoformulations Permit Magnetic Resonance Bioimaging of Antiretroviral Drug Particle Tissue-Cell Biodistribution.
Multimodal Theranostic Nanoformulations Permit Magnetic Resonance Bioimaging of Antiretroviral Drug Particle Tissue-Cell Biodistribution.
Theranostics (2018-01-02)
Bhavesh D Kevadiya, Christopher Woldstad, Brendan M Ottemann, Prasanta Dash, Balasrinivasa R Sajja, Benjamin Lamberty, Brenda Morsey, Ted Kocher, Rinku Dutta, Aditya N Bade, Yutong Liu, Shannon E Callen, Howard S Fox, Siddappa N Byrareddy, JoEllyn M McMillan, Tatiana K Bronich, Benson J Edagwa, Michael D Boska, Howard E Gendelman
ABSTRACT
Long-acting slow effective release antiretroviral therapy (LASER ART) was developed to improve patient regimen adherence, prevent new infections, and facilitate drug delivery to human immunodeficiency virus cell and tissue reservoirs. In an effort to facilitate LASER ART development, "multimodal imaging theranostic nanoprobes" were created. These allow combined bioimaging, drug pharmacokinetics and tissue biodistribution tests in animal models. Europium (Eu Folic acid (FA) decoration of EuCF-DTG (FA-EuCF-DTG) nanoparticles facilitated macrophage targeting and sped drug entry across cell barriers. Macrophage uptake was higher for FA-EuCF-DTG than EuCF-DTG nanoparticles with relaxivities of r We posit that these theranostic nanoprobes can assess LASER ART drug delivery and be used as part of a precision nanomedicine therapeutic strategy.
Sigma-Aldrich
1,3-Di-o-tolylguanidine, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Europium(III) nitrate hydrate, 99.99% trace metals basis
Pricing and availability is not currently available.