Merck
Rescue of ATXN3 neuronal toxicity in
Disease models & mechanisms (2017-10-25)
Yasmin Fardghassemi, Arnaud Tauffenberger, Sarah Gosselin, J Alex Parker
ABSTRACT
Polyglutamine expansion diseases are a group of hereditary neurodegenerative disorders that develop when a CAG repeat in the causative genes is unstably expanded above a certain threshold. The expansion of trinucleotide CAG repeats causes hereditary adult-onset neurodegenerative disorders, such as Huntington's disease, dentatorubral-pallidoluysian atrophy, spinobulbar muscular atrophy and multiple forms of spinocerebellar ataxia (SCA). The most common dominantly inherited SCA is the type 3 (SCA3), also known as Machado-Joseph disease (MJD), which is an autosomal dominant, progressive neurological disorder. The gene causatively associated with MJD is
Sigma-Aldrich
Donkey serum
Pricing and availability is not currently available.