Merck
  • Carbohydrate/DBU Cocatalyzed Alkene Diboration: Mechanistic Insight Provides Enhanced Catalytic Efficiency and Substrate Scope.
Carbohydrate/DBU Cocatalyzed Alkene Diboration: Mechanistic Insight Provides Enhanced Catalytic Efficiency and Substrate Scope.
Journal of the American Chemical Society (2018-02-15)
Lu Yan, Yan Meng, Fredrik Haeffner, Robert M Leon, Michael P Crockett, James P Morken
ABSTRACT
A mechanistic investigation of the carbohydrate/DBU cocatalyzed enantioselective diboration of alkenes is presented. These studies provide an understanding of the origin of stereoselectivity and also reveal a strategy for enhancing reactivity and broadening the substrate scope.
Sigma-Aldrich
2,2′-Bi-1,3,2-dioxaborinane, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
TBS-DHG Catalyst, ≥95%
Pricing and availability is not currently available.