Merck
  • Transdifferentiation of adult rat stem Leydig cells into prostatic and uterine epithelium, but not epidermis.
Transdifferentiation of adult rat stem Leydig cells into prostatic and uterine epithelium, but not epidermis.
Andrology (2017-10-27)
M K Nanjappa, T I Medrano, G S Prins, H Chen, B R Zirkin, P S Cooke
ABSTRACT
Stem Leydig cells (SLCs), precursors of testicular Leydig cells that secrete testosterone required for male sexual differentiation, spermatogenesis, and fertility, were recently identified in rat testes. Various types of stem cells have shown the ability to differentiate into other tissues, but there is no information on the plasticity of adult rat SLCs (rSLCs). This study investigated the ability of rSLCs to transdifferentiate into cell types from all three germ layers-prostatic epithelium (endoderm), uterine epithelium (mesoderm), and epidermis (ectoderm)-under the influence of inductive mesenchyme from fetal and neonatal tissues. To differentiate rSLCs into cells of other lineages, mesenchyme from green fluorescent protein (GFP)-expressing mice was used. Tissue recombinants of urogenital sinus mesenchyme (a potent prostate inducer) and rSLCs grafted into adult male hosts formed ductal structures resembling prostate after 5 weeks. Prostate epithelium was of rSLC origin as determined by absence of GFP expression, and expressed characteristic markers of prostatic epithelium. Similarly, uterine mesenchyme + rSLCs tissue recombinants contained a simple columnar epithelium that was histologically similar to normal uterine epithelium and expressed typical uterine epithelial markers, but was of rSLC origin. In contrast, epidermal tissue was absent in fetal dermis + rSLCs recombinants, suggesting rSLCs did not form skin epithelium. Thus, rSLCs can transdifferentiate into uterine and prostatic epithelium, mesodermal, and endodermal derivatives, respectively, but they may have a limited transdifferentiation potential, as shown by their inability to form epidermis, an ectodermal derivative.
Sigma-Aldrich
Anti-Nkx-3.1 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Basal Medium Eagle, With Earle′s salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Pricing and availability is not currently available.