- Lipid metabolism and pro-oxidant/antioxidant balance of Halamphora oceanica from the Gulf of Mexico exposed to water accommodated fraction of Maya crude oil.
Lipid metabolism and pro-oxidant/antioxidant balance of Halamphora oceanica from the Gulf of Mexico exposed to water accommodated fraction of Maya crude oil.
Diatoms play key roles in primary production and carbon fixation at a global scale and in some cases these species live on marine ecosystems impacted by crude oil (CO) spills. Halamphora oceanica, a new diatom species from the Southwest of the Gulf of Mexico was isolated and cultured in the laboratory and was exposed to water accommodated fraction (WAF) of different Maya CO loads at 0.01, 0.1, 1 and 10g/L by 96h. A battery of biomarkers involved in oxidative stress (O