Merck
  • A Novel AP2/ERF Transcription Factor CR1 Regulates the Accumulation of Vindoline and Serpentine in
A Novel AP2/ERF Transcription Factor CR1 Regulates the Accumulation of Vindoline and Serpentine in
Frontiers in plant science (2017-12-23)
Jiaqi Liu, Fangyuan Gao, Juansheng Ren, Xianjun Lu, Guangjun Ren, Rui Wang
ABSTRACT
As one type of the most important alkaloids in the world, terpenoid indole alkaloids (TIAs) show a wide range of pharmaceutical activities that are beneficial for clinical treatments.